| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
ENST00000236273.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
ENST00000236273.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
ENST00000236273.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
ENST00000236273.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
ENST00000236273.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
ENST00000236273.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
ENST00000236273.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
ENST00000236273.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
ENST00000354361.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
ENST00000354361.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
ENST00000354361.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
ENST00000354361.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
ENST00000354361.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
ENST00000354361.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
ENST00000474160.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
ENST00000474160.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
ENST00000474160.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
ENST00000476231.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
ENST00000476231.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
NM_015484.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
NM_015484.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
NM_015484.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
NM_015484.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
NM_015484.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
NM_015484.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
NM_015484.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
NM_015484.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
NM_015484.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
NM_207170.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
NM_207170.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
NM_207170.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
NM_207170.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
NM_207170.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
NM_207170.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
NM_207170.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100013348.hg.1 |
NM_207170.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01036579.hg.1 |
ENST00000236273.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01036579.hg.1 |
ENST00000354361.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01036579.hg.1 |
NM_015484.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01036579.hg.1 |
NM_207170.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01036579.hg.1 |
TC01002373.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
ENST00000236273.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
ENST00000236273.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
ENST00000236273.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
ENST00000236273.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
ENST00000236273.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
ENST00000236273.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
ENST00000236273.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
ENST00000236273.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
ENST00000354361.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
ENST00000354361.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
ENST00000354361.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
ENST00000354361.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
ENST00000354361.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
ENST00000354361.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
ENST00000474160.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
ENST00000474160.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
ENST00000474160.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
ENST00000476231.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
ENST00000476231.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
NM_015484.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
NM_015484.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
NM_015484.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
NM_015484.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
NM_015484.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
NM_015484.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
NM_015484.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
NM_015484.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
NM_015484.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
NM_207170.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
NM_207170.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
NM_207170.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
NM_207170.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
NM_207170.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
NM_207170.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
NM_207170.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100013348.hg.1 |
NM_207170.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199061.hg.1 |
ENST00000236273.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199061.hg.1 |
ENST00000354361.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199061.hg.1 |
NM_015484.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199061.hg.1 |
NM_207170.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199061.hg.1 |
ENST00000236273.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199061.hg.1 |
ENST00000354361.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100199061.hg.1 |
NM_015484.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199061.hg.1 |
NM_207170.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100199061.hg.1 |
TC0100013348.hg.1 |
Clariom D TC |