This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01549303_m1
See other TGDS GE Assays ›
Gene Symbol:
TGDS
Gene Name:
TDP-glucose 4,6-dehydratase
Gene Aliases:
CATMANS, SDR2E1, TDPGD
Chromosome Location:
Chr.13: 94574054 - 94596275 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01549303_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TGDS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001304430.1 NP_001291359.1 6 - 7 761 143
NM_014305.3 NP_055120.1 6 - 7 685 143
NR_130731.1 6 - 7 681 143
NR_130732.1 5 - 6 594 143
XM_011521065.2 XP_011519367.1 6 - 7 688 143
XM_011521066.2 XP_011519368.1 6 - 7 569 143
GenBank mRNA AF048686.1 - 6 - 7 587 143
AJ006068.1 - 6 - 7 639 143
AK313251.1 - 6 - 7 595 143
AY544125.1 - 6 - 7 562 143
BC005284.1 - 6 - 7 667 143
BC033675.1 - 6 - 7 614 5175390 143
BG702561.1 - 6 - 7 571 144

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300009492.hg.1
This assay targets the same location as HTA transcript cluster(s) TC13000793.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TGDS

Panther Classification:

Molecular Function -

dehydratase

Gene Ontology Categories:

Function(s) Process(es)

nucleotide-sugar metabolic process
dTDP-glucose 4,6-dehydratase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR13008905.hg.1 ENST00000261296.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13008905.hg.1 ENST00000498294.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13008905.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13008905.hg.1 TC13000793.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000470480.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000470480.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000470480.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay PSR1300163508.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay PSR1300163508.hg.1 ENST00000470480.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300163508.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300163508.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay PSR1300163508.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay PSR1300163508.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay PSR1300163508.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay PSR1300163508.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay PSR1300163508.hg.1 ENST00000470480.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300163508.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Clariom D Assay PSR1300163508.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay PSR1300163508.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay PSR1300163508.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay PSR1300163508.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay PSR1300163508.hg.1 TC1300009492.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000470480.1 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000470480.1 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000470480.1 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130732.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0