This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01549305_g1
See other TGDS GE Assays ›
Gene Symbol:
TGDS
Gene Name:
TDP-glucose 4,6-dehydratase
Gene Aliases:
CATMANS, SDR2E1, TDPGD
Chromosome Location:
Chr.13: 94574054 - 94596275 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01549305_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TGDS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001304430.1 NP_001291359.1 8 - 9 879 147
NM_014305.3 NP_055120.1 8 - 9 803 147
NR_130731.1 8 - 9 799 147
NR_130732.1 7 - 8 712 147
XM_011521065.2 XP_011519367.1 8 - 9 806 147
XM_011521066.2 XP_011519368.1 8 - 9 687 147
GenBank mRNA AF048686.1 - 8 - 9 705 147
AJ006068.1 - 8 - 9 757 147
AK313251.1 - 8 - 9 713 147
AY544125.1 - 8 - 9 680 147
BC005284.1 - 8 - 9 785 147
BC033675.1 - 8 - 9 732 5175390 147

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300009492.hg.1
This assay targets the same location as HTA transcript cluster(s) TC13000793.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TGDS

Panther Classification:

Molecular Function -

dehydratase

Gene Ontology Categories:

Function(s) Process(es)

nucleotide-sugar metabolic process
dTDP-glucose 4,6-dehydratase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000470480.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000470480.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000470480.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13008902.hg.1 ENST00000261296.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR13008902.hg.1 NM_014305.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR13008902.hg.1 TC13000793.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000470480.1 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000470480.1 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000470480.1 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 ENST00000498294.5 Ensembl
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay TC1300009492.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay PSR1300163505.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay PSR1300163505.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay PSR1300163505.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay PSR1300163505.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay PSR1300163505.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay PSR1300163505.hg.1 ENST00000261296.6 Ensembl
Affymetrix Human Clariom D Assay PSR1300163505.hg.1 NM_001304430.1 Refseq
Affymetrix Human Clariom D Assay PSR1300163505.hg.1 NM_014305.3 Refseq
Affymetrix Human Clariom D Assay PSR1300163505.hg.1 NR_130731.1 Refseq
Affymetrix Human Clariom D Assay PSR1300163505.hg.1 NR_130732.1 Refseq
Affymetrix Human Clariom D Assay PSR1300163505.hg.1 TC1300009492.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0