This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01549515_m1
See other CADM2 GE Assays ›
Gene Symbol:
CADM2
Gene Name:
cell adhesion molecule 2
Gene Aliases:
IGSF4D, NECL3, Necl-3, SynCAM 2, synCAM2
Chromosome Location:
Chr.3: 84958982 - 86074429 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01549515_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CADM2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001167674.1 NP_001161146.1 8 - 9 1687 96
NM_001256502.1 NP_001243431.1 9 - 10 1234 96
NM_001256503.1 NP_001243432.1 8 - 9 1084 96
NM_153184.3 NP_694854.2 8 - 9 1067 96
XM_006713081.3 XP_006713144.1 9 - 10 1577 96
XM_017006062.1 XP_016861551.1 10 - 11 1619 96
GenBank mRNA AB646741.1 - 9 - 10 1234 96
AB646743.1 - 8 - 9 1084 96
AF538973.1 - 8 - 9 1067 96
AL834270.1 - 8 - 9 1657 96
BC112136.1 - 8 - 9 1123 8327604 96
BC112138.1 - 8 - 9 1135 8327606 96

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300007973.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03000455.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CADM2

Gene Ontology Categories:

Function(s) Process(es)

homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
cell recognition
adherens junction organization
receptor activity
receptor binding
protein homodimerization activity
cell adhesion molecule binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR03008994.hg.1 TC03000455.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03008994.hg.1 ENST00000383699.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03008994.hg.1 ENST00000405615.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03008994.hg.1 ENST00000407528.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03008994.hg.1 NM_001167674.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03008994.hg.1 NM_001167675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03008994.hg.1 NM_153184.3 Refseq
Affymetrix Human Clariom D Assay PSR0300157034.hg.1 ENST00000383699.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300157034.hg.1 ENST00000405615.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300157034.hg.1 ENST00000407528.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300157034.hg.1 NM_001167674.1 Refseq
Affymetrix Human Clariom D Assay PSR0300157034.hg.1 NM_001167675.1 Refseq
Affymetrix Human Clariom D Assay PSR0300157034.hg.1 NM_001256502.1 Refseq
Affymetrix Human Clariom D Assay PSR0300157034.hg.1 NM_001256503.1 Refseq
Affymetrix Human Clariom D Assay PSR0300157034.hg.1 NM_001256504.1 Refseq
Affymetrix Human Clariom D Assay PSR0300157034.hg.1 NM_001256505.1 Refseq
Affymetrix Human Clariom D Assay PSR0300157034.hg.1 NM_153184.3 Refseq
Affymetrix Human Clariom D Assay PSR0300157034.hg.1 ENST00000383699.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300157034.hg.1 ENST00000405615.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300157034.hg.1 ENST00000407528.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300157034.hg.1 NM_001167674.1 Refseq
Affymetrix Human Clariom D Assay PSR0300157034.hg.1 NM_001167675.1 Refseq
Affymetrix Human Clariom D Assay PSR0300157034.hg.1 NM_001256502.1 Refseq
Affymetrix Human Clariom D Assay PSR0300157034.hg.1 NM_001256503.1 Refseq
Affymetrix Human Clariom D Assay PSR0300157034.hg.1 NM_001256504.1 Refseq
Affymetrix Human Clariom D Assay PSR0300157034.hg.1 NM_001256505.1 Refseq
Affymetrix Human Clariom D Assay PSR0300157034.hg.1 NM_153184.3 Refseq
Affymetrix Human Clariom D Assay PSR0300157034.hg.1 TC0300007973.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0