This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01550240_m1
See other ADGRG7 GE Assays ›
Gene Symbol:
ADGRG7
Gene Name:
adhesion G protein-coupled receptor G7
Gene Aliases:
GPR128
Chromosome Location:
Chr.3: 100609589 - 100696174 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01550240_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADGRG7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_032787.2 NP_116176.2 6 - 7 960 92
XM_011513245.2 XP_011511547.1 5 - 6 835 92
GenBank mRNA AK027360.1 - 6 - 7 948 92
AK172769.1 - 2 - 3 202 92
AY181246.1 - 6 - 7 692 92
BC113451.1 - 6 - 7 726 8322503 92
BC130388.1 - 6 - 7 726 40146419 92
DA640995.1 - 2 - 3 386 92

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300008150.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03000510.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK172769.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ADGRG7

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
G-protein coupled receptor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR03009604.hg.1 TC03000510.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03009604.hg.1 ENST00000273352.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03009604.hg.1 NM_032787.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000475887.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000475887.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000475887.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000475887.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000475887.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000475887.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000475887.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000475887.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000475887.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000475887.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000475887.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000475887.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000475887.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000481361.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000481361.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000481361.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000481361.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000481361.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000481361.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000481506.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000481506.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000481506.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000481506.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000481506.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000481506.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000481506.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000481506.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000481506.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000493081.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000493081.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000493081.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000493081.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 ENST00000493081.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000475887.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000475887.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000475887.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000475887.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000475887.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000475887.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000475887.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000475887.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000475887.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000475887.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000475887.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000475887.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000475887.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000481361.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000481361.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000481361.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000481361.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000481361.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000481361.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000481506.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000481506.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000481506.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000481506.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000481506.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000481506.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000481506.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000481506.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000481506.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000493081.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000493081.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000493081.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000493081.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 ENST00000493081.1 Ensembl
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_001308362.1 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Clariom D Assay TC0300008150.hg.1 NM_032787.2 Refseq
Affymetrix Human Clariom D Assay PSR0300158103.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300158103.hg.1 ENST00000481361.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300158103.hg.1 NM_032787.2 Refseq
Affymetrix Human Clariom D Assay PSR0300158103.hg.1 ENST00000273352.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300158103.hg.1 ENST00000481361.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300158103.hg.1 NM_032787.2 Refseq
Affymetrix Human Clariom D Assay PSR0300158103.hg.1 TC0300008150.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0