This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01550446_m1
See other NME6 GE Assays ›
Gene Symbol:
NME6
Gene Name:
NME/NM23 nucleoside diphosphate kinase 6
Gene Aliases:
IPIA-ALPHA, NDK 6, NM23-H6
Chromosome Location:
Chr.3: 48289369 - 48302259 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01550446_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NME6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001308426.1 NP_001295355.1 2 - 3 426 94
NM_001308427.1 NP_001295356.1 2 - 3 235 94
NM_001308428.1 NP_001295357.1 2 - 3 226 94
NM_001308430.1 NP_001295359.1 2 - 3 209 94
NM_001308431.1 NP_001295360.1 2 - 3 426 94
NM_001308433.1 NP_001295362.1 2 - 3 226 94
NM_001308435.1 NP_001295364.1 2 - 3 426 94
NM_005793.4 NP_005784.1 2 - 3 209 94
XM_017005511.1 XP_016861000.1 2 - 3 137 94
XM_017005513.1 XP_016861002.1 2 - 3 151 94
XM_017005514.1 XP_016861003.1 3 - 4 408 94
XM_017005515.1 XP_016861004.1 2 - 3 204 94
XM_017005516.1 XP_016861005.1 2 - 3 166 94
GenBank mRNA AF051941.1 - 2 - 3 142 94
AK222555.1 - 1 - 2 108 94
AK294809.1 - 2 - 3 182 94
AK297364.1 - 2 - 3 135 94
BC001808.2 - 2 - 3 156 2966629 94
BC012828.2 - 2 - 3 156 2966629 94
BE866839.1 - 2 - 3 177 94
BG284082.1 - 2 - 3 155 94
CA848473.1 - 2 - 3 205 94
CD701581.1 - 2 - 3 179 94
CT002144.1 - 2 - 3 426 94
DA398008.1 - 2 - 3 182 94
U90449.1 - 2 - 3 156 94

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NME6

Panther Classification:

Molecular Function -

kinase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

nucleoside diphosphate phosphorylation
GTP biosynthetic process
UTP biosynthetic process
CTP biosynthetic process
apoptotic process
negative regulation of cell growth
negative regulation of mitotic nuclear division
nucleoside diphosphate kinase activity
ATP binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline