This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01550739_m1
See other PCMTD1 GE Assays ›
Gene Symbol:
PCMTD1
Gene Name:
protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.8: 51817575 - 51899186 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01550739_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PCMTD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_052937.3 NP_443169.2 3 - 4 763 105
XM_005251146.2 XP_005251203.1 3 - 4 763 105
XM_011517444.2 XP_011515746.1 2 - 3 352 105
GenBank mRNA AK056952.1 - 3 - 4 613 105
AL833027.1 - 2 - 3 164 105
BC032670.1 - 2 - 3 508 5539318 105
BX640885.1 - 1 - 2 73 105

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0800010417.hg.1
This assay targets the same location as HTA transcript cluster(s) TC08001205.hg.1

Additional Information:

This assay is in the 5'UTR region of the XM_011517444.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PCMTD1

Panther Classification:

Molecular Function -

methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

protein methylation
protein-L-isoaspartate (D-aspartate) O-methyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR08015497.hg.1 TC08001205.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR08015497.hg.1 ENST00000360540.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08015497.hg.1 ENST00000519559.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08015497.hg.1 ENST00000522514.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08015497.hg.1 ENST00000544451.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08015497.hg.1 NM_052937.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000360540.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000360540.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000360540.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000360540.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000360540.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000360540.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000360540.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000360540.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000360540.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000360540.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000360540.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000360540.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000519554.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000519554.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000519554.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000519554.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000519559.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000519559.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000519559.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000519559.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000519559.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000519559.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000519559.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000519975.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000519975.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000521046.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000521046.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000521046.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000521046.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000521046.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000521344.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000521344.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000521344.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000521344.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000521344.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000521344.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000522514.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000522514.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000522514.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000522514.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000522514.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000522514.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000522514.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000522514.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000544451.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000544451.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000544451.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000544451.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000544451.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000544451.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000544451.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000544451.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 ENST00000544451.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_001286782.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_001286782.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_001286782.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_001286782.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_001286782.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_001286782.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_001286782.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_001286782.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_001286782.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_001286783.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_001286783.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_001286783.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_001286783.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_001286783.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_001286783.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_001286783.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_001286783.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_001286783.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_001286783.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_001286783.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_052937.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_052937.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_052937.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_052937.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_052937.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_052937.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_052937.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_052937.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_052937.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_052937.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_052937.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800010417.hg.1 NM_052937.3 Refseq
Affymetrix Human Clariom D Assay PSR0800170698.hg.1 ENST00000360540.9 Ensembl
Affymetrix Human Clariom D Assay PSR0800170698.hg.1 ENST00000519554.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800170698.hg.1 ENST00000519559.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800170698.hg.1 ENST00000522514.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800170698.hg.1 ENST00000544451.2 Ensembl
Affymetrix Human Clariom D Assay PSR0800170698.hg.1 NM_001286782.1 Refseq
Affymetrix Human Clariom D Assay PSR0800170698.hg.1 NM_001286783.1 Refseq
Affymetrix Human Clariom D Assay PSR0800170698.hg.1 NM_052937.3 Refseq
Affymetrix Human Clariom D Assay PSR0800170698.hg.1 ENST00000360540.9 Ensembl
Affymetrix Human Clariom D Assay PSR0800170698.hg.1 ENST00000519554.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800170698.hg.1 ENST00000519559.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800170698.hg.1 ENST00000522514.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800170698.hg.1 ENST00000544451.2 Ensembl
Affymetrix Human Clariom D Assay PSR0800170698.hg.1 NM_001286782.1 Refseq
Affymetrix Human Clariom D Assay PSR0800170698.hg.1 NM_001286783.1 Refseq
Affymetrix Human Clariom D Assay PSR0800170698.hg.1 NM_052937.3 Refseq
Affymetrix Human Clariom D Assay PSR0800170698.hg.1 TC0800010417.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000360540.9 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000360540.9 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000360540.9 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000360540.9 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000360540.9 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000360540.9 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000360540.9 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000360540.9 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000360540.9 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000360540.9 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000360540.9 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000360540.9 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000519554.5 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000519554.5 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000519554.5 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000519554.5 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000519559.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000519559.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000519559.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000519559.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000519559.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000519559.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000519559.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000519975.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000519975.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000521046.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000521046.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000521046.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000521046.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000521046.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000521344.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000521344.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000521344.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000521344.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000521344.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000521344.1 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000522514.5 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000522514.5 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000522514.5 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000522514.5 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000522514.5 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000522514.5 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000522514.5 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000522514.5 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000544451.2 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000544451.2 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000544451.2 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000544451.2 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000544451.2 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000544451.2 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000544451.2 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000544451.2 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 ENST00000544451.2 Ensembl
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_001286782.1 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_001286782.1 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_001286782.1 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_001286782.1 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_001286782.1 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_001286782.1 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_001286782.1 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_001286782.1 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_001286782.1 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_001286783.1 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_001286783.1 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_001286783.1 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_001286783.1 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_001286783.1 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_001286783.1 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_001286783.1 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_001286783.1 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_001286783.1 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_001286783.1 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_001286783.1 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_052937.3 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_052937.3 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_052937.3 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_052937.3 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_052937.3 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_052937.3 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_052937.3 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_052937.3 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_052937.3 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_052937.3 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_052937.3 Refseq
Affymetrix Human Clariom D Assay TC0800010417.hg.1 NM_052937.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline