This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01551541_m1
See other PTPRD GE Assays ›
Gene Symbol:
PTPRD
Gene Name:
protein tyrosine phosphatase, receptor type D
Gene Aliases:
HPTP, HPTPD, HPTPDELTA, PTPD, RPTPDELTA
Chromosome Location:
Chr.9: 8314246 - 10612840 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01551541_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PTPRD
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002839.3 NP_002830.1 26 - 27 3098 85
XM_006716817.3 XP_006716880.1 25 - 26 3396 85
XM_006716823.2 XP_006716886.1 25 - 26 3390 85
XM_006716825.3 XP_006716888.1 25 - 26 3392 85
XM_011517992.2 XP_011516294.1 25 - 26 3389 85
XM_017014958.1 XP_016870447.1 25 - 26 3395 85
XM_017014959.1 XP_016870448.1 26 - 27 3467 85
XM_017014960.1 XP_016870449.1 24 - 25 3356 85
XM_017014961.1 XP_016870450.1 24 - 25 3340 85
XM_017014962.1 XP_016870451.1 16 - 17 2528 85
XM_017014963.1 XP_016870452.1 25 - 26 3395 85
XM_017014964.1 XP_016870453.1 25 - 26 3394 85
XM_017014965.1 XP_016870454.1 25 - 26 3397 85
XM_017014966.1 XP_016870455.1 24 - 25 3386 85
XM_017014967.1 XP_016870456.1 24 - 25 3381 85
XM_017014968.1 XP_016870457.1 25 - 26 3392 85
XM_017014969.1 XP_016870458.1 24 - 25 3377 85
XM_017014970.1 XP_016870459.1 25 - 26 3395 85
XM_017014971.1 XP_016870460.1 23 - 24 3366 85
XM_017014973.1 XP_016870462.1 23 - 24 3362 85
XM_017014975.1 XP_016870464.1 22 - 23 3351 85
XM_017014976.1 XP_016870465.1 25 - 26 3392 85
XM_017014978.1 XP_016870467.1 25 - 26 3392 85
XM_017014979.1 XP_016870468.1 25 - 26 3394 85
GenBank mRNA AB211400.1 - 25 - 26 3066 85
L38929.1 - 16 - 17 2508 85
X54133.1 - 6 - 7 1188 85

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900009520.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09000906.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Phosphatase Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PTPRD

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
phosphate-containing compound metabolic process
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
transmembrane receptor protein tyrosine phosphatase signaling pathway
chemical synaptic transmission
neuron differentiation
peptidyl-tyrosine dephosphorylation
positive regulation of dendrite morphogenesis
presynaptic membrane assembly
transmembrane receptor protein tyrosine phosphatase activity
receptor binding
protein binding
cell adhesion molecule binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR09012024.hg.1 ENST00000356435.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09012024.hg.1 ENST00000358503.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09012024.hg.1 ENST00000360074.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09012024.hg.1 ENST00000381196.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09012024.hg.1 ENST00000471274.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09012024.hg.1 ENST00000540109.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09012024.hg.1 NM_002839.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09012024.hg.1 TC09000906.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0900165243.hg.1 ENST00000356435.9 Ensembl
Affymetrix Human Clariom D Assay PSR0900165243.hg.1 ENST00000381196.8 Ensembl
Affymetrix Human Clariom D Assay PSR0900165243.hg.1 ENST00000471274.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900165243.hg.1 ENST00000540109.5 Ensembl
Affymetrix Human Clariom D Assay PSR0900165243.hg.1 NM_002839.3 Refseq
Affymetrix Human Clariom D Assay PSR0900165243.hg.1 ENST00000356435.9 Ensembl
Affymetrix Human Clariom D Assay PSR0900165243.hg.1 ENST00000381196.8 Ensembl
Affymetrix Human Clariom D Assay PSR0900165243.hg.1 ENST00000471274.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900165243.hg.1 ENST00000540109.5 Ensembl
Affymetrix Human Clariom D Assay PSR0900165243.hg.1 NM_002839.3 Refseq
Affymetrix Human Clariom D Assay PSR0900165243.hg.1 TC0900009520.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline