This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01551821_m1
See other MRI1 GE Assays ›
Gene Symbol:
MRI1
Gene Name:
methylthioribose-1-phosphate isomerase 1
Gene Aliases:
MRDI, MTNA, Ypr118w
Chromosome Location:
Chr.19: 13764492 - 13774282 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01551821_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MRI1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001031727.2 NP_001026897.1 2 - 3 442 74
XM_011528356.2 XP_011526658.1 2 - 3 465 74
XM_011528357.2 XP_011526659.1 2 - 3 473 74
GenBank mRNA AY358176.1 - 2 - 3 419 74
BC001703.1 - 2 - 3 416 3501899 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900011674.hg.1
This assay targets the same location as HTA transcript cluster(s) TC19000239.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MRI1

Panther Classification:

Molecular Function -

isomerase

Gene Ontology Categories:

Function(s) Process(es)

L-methionine biosynthetic process from S-adenosylmethionine
L-methionine biosynthetic process from methylthioadenosine
identical protein binding
S-methyl-5-thioribose-1-phosphate isomerase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR19003534.hg.1 ENST00000040663.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19003534.hg.1 NM_001031727.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19003534.hg.1 TC19000239.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000040663.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000040663.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000040663.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000040663.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000040663.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000040663.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000040663.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000040663.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000040663.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000040663.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000040663.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000040663.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000319545.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000319545.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000319545.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000319545.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000319545.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000319545.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000319545.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000319545.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000319545.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000319545.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000319545.12 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000588526.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000588526.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000588526.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000588526.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000588526.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000588526.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000588526.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000588526.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000589290.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000589290.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000589290.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000589290.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000591688.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000591688.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000591688.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000591688.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000591688.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000591688.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000591688.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000591688.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000591688.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000591688.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000593245.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000593245.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000593245.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000593245.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000593245.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000593245.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 ENST00000593245.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 NM_001031727.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 NM_001031727.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 NM_001031727.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 NM_001031727.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 NM_001031727.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 NM_001031727.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 NM_001031727.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 NM_001031727.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 NM_001031727.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 NM_001031727.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 NM_001031727.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 NM_001031727.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 NM_001031727.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 NM_032285.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 NM_032285.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 NM_032285.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 NM_032285.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 NM_032285.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 NM_032285.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 NM_032285.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 NM_032285.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 NM_032285.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 NM_032285.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011674.hg.1 NM_032285.3 Refseq
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000040663.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000040663.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000040663.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000040663.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000040663.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000040663.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000040663.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000040663.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000040663.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000040663.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000040663.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000040663.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000319545.12 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000319545.12 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000319545.12 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000319545.12 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000319545.12 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000319545.12 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000319545.12 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000319545.12 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000319545.12 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000319545.12 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000319545.12 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000588526.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000588526.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000588526.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000588526.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000588526.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000588526.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000588526.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000588526.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000589290.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000589290.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000589290.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000589290.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000591688.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000591688.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000591688.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000591688.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000591688.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000591688.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000591688.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000591688.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000591688.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000591688.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000593245.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000593245.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000593245.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000593245.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000593245.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000593245.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 ENST00000593245.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011674.hg.1 NM_001031727.3 Refseq
Affymetrix Human Clariom D Assay TC1900011674.hg.1 NM_001031727.3 Refseq
Affymetrix Human Clariom D Assay TC1900011674.hg.1 NM_001031727.3 Refseq
Affymetrix Human Clariom D Assay TC1900011674.hg.1 NM_001031727.3 Refseq
Affymetrix Human Clariom D Assay TC1900011674.hg.1 NM_001031727.3 Refseq
Affymetrix Human Clariom D Assay TC1900011674.hg.1 NM_001031727.3 Refseq
Affymetrix Human Clariom D Assay TC1900011674.hg.1 NM_001031727.3 Refseq
Affymetrix Human Clariom D Assay TC1900011674.hg.1 NM_001031727.3 Refseq
Affymetrix Human Clariom D Assay TC1900011674.hg.1 NM_001031727.3 Refseq
Affymetrix Human Clariom D Assay TC1900011674.hg.1 NM_001031727.3 Refseq
Affymetrix Human Clariom D Assay TC1900011674.hg.1 NM_001031727.3 Refseq
Affymetrix Human Clariom D Assay TC1900011674.hg.1 NM_001031727.3 Refseq
Affymetrix Human Clariom D Assay TC1900011674.hg.1 NM_001031727.3 Refseq
Affymetrix Human Clariom D Assay TC1900011674.hg.1 NM_032285.3 Refseq
Affymetrix Human Clariom D Assay TC1900011674.hg.1 NM_032285.3 Refseq
Affymetrix Human Clariom D Assay TC1900011674.hg.1 NM_032285.3 Refseq
Affymetrix Human Clariom D Assay TC1900011674.hg.1 NM_032285.3 Refseq
Affymetrix Human Clariom D Assay TC1900011674.hg.1 NM_032285.3 Refseq
Affymetrix Human Clariom D Assay TC1900011674.hg.1 NM_032285.3 Refseq
Affymetrix Human Clariom D Assay TC1900011674.hg.1 NM_032285.3 Refseq
Affymetrix Human Clariom D Assay TC1900011674.hg.1 NM_032285.3 Refseq
Affymetrix Human Clariom D Assay TC1900011674.hg.1 NM_032285.3 Refseq
Affymetrix Human Clariom D Assay TC1900011674.hg.1 NM_032285.3 Refseq
Affymetrix Human Clariom D Assay TC1900011674.hg.1 NM_032285.3 Refseq
Affymetrix Human Clariom D Assay PSR1900199542.hg.1 ENST00000040663.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900199542.hg.1 ENST00000588526.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900199542.hg.1 ENST00000591688.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900199542.hg.1 ENST00000593245.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900199542.hg.1 NM_001031727.3 Refseq
Affymetrix Human Clariom D Assay PSR1900199542.hg.1 TC1900011674.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900199542.hg.1 ENST00000040663.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900199542.hg.1 ENST00000588526.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900199542.hg.1 ENST00000591688.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900199542.hg.1 ENST00000593245.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900199542.hg.1 NM_001031727.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline