This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01551886_m1
See other ALDOB GE Assays ›
Gene Symbol:
ALDOB
Gene Name:
aldolase, fructose-bisphosphate B
Gene Aliases:
ALDB, ALDO2
Chromosome Location:
Chr.9: 101420560 - 101435780 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01551886_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALDOB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000035.3 NP_000026.2 2 - 3 190 75
GenBank mRNA AK290795.1 - 2 - 3 184 75
BC029399.1 - 2 - 3 184 4593670 75
X01098.1 - 2 - 3 174 75
X02747.1 - 2 - 3 233 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900011033.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09001421.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ALDOB

Panther Classification:

Molecular Function -

aldolase

Gene Ontology Categories:

Function(s) Process(es)

liver development
fructose metabolic process
gluconeogenesis
glycolytic process
NADH oxidation
response to fructose
response to zinc ion
fructose 1,6-bisphosphate metabolic process
cellular response to extracellular stimulus
positive regulation of ATPase activity
cellular response to insulin stimulus
response to drug
response to starvation
response to amino acid
response to copper ion
response to glucocorticoid
response to cAMP
canonical glycolysis
fructose catabolic process to hydroxyacetone phosphate and glyceraldehyde-3-phosphate
vacuolar proton-transporting V-type ATPase complex assembly
response to interleukin-6
fructose-bisphosphate aldolase activity
protein binding
cytoskeletal protein binding
phosphatidylcholine binding
identical protein binding
ATPase binding
fructose-1-phosphate aldolase activity
fructose binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000468981.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000468981.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000468981.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09017742.hg.1 TC09001421.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09017742.hg.1 ENST00000374855.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09017742.hg.1 ENST00000430164.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09017742.hg.1 NM_000035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09017741.hg.1 TC09001421.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09017741.hg.1 ENST00000374855.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09017741.hg.1 ENST00000430164.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09017741.hg.1 ENST00000468981.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09017741.hg.1 NM_000035.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09017743.hg.1 TC09001421.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09017743.hg.1 ENST00000374855.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09017743.hg.1 ENST00000430164.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09017743.hg.1 ENST00000468981.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09017743.hg.1 NM_000035.3 Refseq
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000468981.3 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000468981.3 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000468981.3 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Clariom D Assay TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Clariom D Assay TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Clariom D Assay TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Clariom D Assay TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Clariom D Assay TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Clariom D Assay TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Clariom D Assay TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Clariom D Assay TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Clariom D Assay TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Clariom D Assay TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Clariom D Assay TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Clariom D Assay TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Clariom D Assay TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Clariom D Assay TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Clariom D Assay TC0900011033.hg.1 NM_000035.3 Refseq
Affymetrix Human Clariom D Assay PSR0900175206.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Clariom D Assay PSR0900175206.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900175206.hg.1 NM_000035.3 Refseq
Affymetrix Human Clariom D Assay PSR0900175206.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Clariom D Assay PSR0900175206.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900175206.hg.1 NM_000035.3 Refseq
Affymetrix Human Clariom D Assay PSR0900175206.hg.1 TC0900011033.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0900175205.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Clariom D Assay PSR0900175205.hg.1 ENST00000468981.3 Ensembl
Affymetrix Human Clariom D Assay PSR0900175205.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900175205.hg.1 NM_000035.3 Refseq
Affymetrix Human Clariom D Assay PSR0900175205.hg.1 ENST00000374855.8 Ensembl
Affymetrix Human Clariom D Assay PSR0900175205.hg.1 ENST00000468981.3 Ensembl
Affymetrix Human Clariom D Assay PSR0900175205.hg.1 ENST00000616752.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900175205.hg.1 NM_000035.3 Refseq
Affymetrix Human Clariom D Assay PSR0900175205.hg.1 TC0900011033.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0