This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01552664_g1
See other NDUFAF3 GE Assays ›
Gene Symbol:
NDUFAF3
Gene Name:
NADH:ubiquinone oxidoreductase complex assembly factor 3
Gene Aliases:
2P1, C3orf60, E3-3
Chromosome Location:
Chr.3: 49020475 - 49023495 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01552664_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NDUFAF3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199070.1 NP_951033.1 1 - 2 142 91
GenBank mRNA AL049955.1 - 1 - 2 146 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300007359.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03000287.hg.1

Additional Information:

This assay is in the 5'UTR region of the AL049955.1,NM_199070.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NDUFAF3

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial respiratory chain complex I assembly
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000326912.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000326912.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000326912.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000326912.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000326912.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000326912.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000326912.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000326925.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000326925.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000326925.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000326925.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000326925.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000326925.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000326925.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000326925.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000326925.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000326925.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000326925.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000326925.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000326925.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000326925.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000395458.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000395458.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000395458.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000395458.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000395458.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000395458.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000395458.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000451378.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000451378.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000451378.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000451378.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000451378.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000451378.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000451378.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000451378.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000480392.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000480392.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000480392.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000480392.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000480392.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000480392.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000480392.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000480392.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000496152.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000496152.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000496152.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000496152.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000496152.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000496152.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000496152.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 ENST00000496152.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199069.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199069.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199069.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199069.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199069.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199069.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199069.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199069.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199069.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199069.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199070.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199070.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199070.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199070.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199070.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199070.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199070.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199070.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199073.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199073.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199073.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199073.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199073.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199073.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199073.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199073.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199074.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199074.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199074.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199074.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199074.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199074.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199074.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300007359.hg.1 NM_199074.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03005893.hg.1 TC03000287.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03005893.hg.1 ENST00000326925.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03005893.hg.1 ENST00000451378.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03005893.hg.1 NM_199069.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03005893.hg.1 NM_199070.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000326912.8 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000326912.8 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000326912.8 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000326912.8 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000326912.8 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000326912.8 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000326912.8 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000326925.10 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000326925.10 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000326925.10 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000326925.10 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000326925.10 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000326925.10 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000326925.10 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000326925.10 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000326925.10 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000326925.10 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000326925.10 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000326925.10 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000326925.10 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000326925.10 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000395458.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000395458.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000395458.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000395458.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000395458.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000395458.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000395458.6 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000451378.2 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000451378.2 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000451378.2 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000451378.2 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000451378.2 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000451378.2 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000451378.2 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000451378.2 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000480392.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000480392.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000480392.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000480392.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000480392.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000480392.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000480392.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000480392.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000496152.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000496152.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000496152.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000496152.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000496152.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000496152.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000496152.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 ENST00000496152.1 Ensembl
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199069.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199069.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199069.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199069.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199069.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199069.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199069.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199069.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199069.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199069.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199070.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199070.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199070.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199070.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199070.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199070.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199070.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199070.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199073.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199073.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199073.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199073.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199073.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199073.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199073.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199073.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199074.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199074.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199074.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199074.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199074.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199074.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199074.1 Refseq
Affymetrix Human Clariom D Assay TC0300007359.hg.1 NM_199074.1 Refseq
Affymetrix Human Clariom D Assay PSR0300152279.hg.1 ENST00000326925.10 Ensembl
Affymetrix Human Clariom D Assay PSR0300152279.hg.1 ENST00000451378.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300152279.hg.1 NM_199069.1 Refseq
Affymetrix Human Clariom D Assay PSR0300152279.hg.1 NM_199070.1 Refseq
Affymetrix Human Clariom D Assay PSR0300152279.hg.1 ENST00000326925.10 Ensembl
Affymetrix Human Clariom D Assay PSR0300152279.hg.1 ENST00000451378.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300152279.hg.1 NM_199069.1 Refseq
Affymetrix Human Clariom D Assay PSR0300152279.hg.1 NM_199070.1 Refseq
Affymetrix Human Clariom D Assay PSR0300152279.hg.1 TC0300007359.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0