This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01552924_m1
See other FGFR2 GE Assays ›
Gene Symbol:
FGFR2
Gene Name:
fibroblast growth factor receptor 2
Gene Aliases:
BBDS, BEK, BFR-1, CD332, CEK3, CFD1, ECT1, JWS, K-SAM, KGFR, TK14, TK25
Chromosome Location:
Chr.10: 121478330 - 121598656 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01552924_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FGFR2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000141.4 NP_000132.3 3 - 4 1022 84
NM_001144913.1 NP_001138385.1 2 - 3 525 84
NM_001144914.1 NP_001138386.1 2 - 3 525 84
NM_001144917.1 NP_001138389.1 3 - 4 1022 84
NM_001320658.1 NP_001307587.1 3 - 4 1022 84
NM_022970.3 NP_075259.4 3 - 4 1022 84
XM_006717708.2 XP_006717771.1 3 - 4 1220 84
XM_006717710.3 XP_006717773.1 3 - 4 1220 84
XM_017015920.1 XP_016871409.1 3 - 4 1220 84
XM_017015921.1 XP_016871410.1 3 - 4 1220 84
GenBank mRNA AB030073.1 - 3 - 4 793 84
AB030074.1 - 3 - 4 793 84
AB030075.1 - 3 - 4 793 84
AB030076.1 - 3 - 4 793 84
AB030077.1 - 3 - 4 793 84
AB030078.1 - 3 - 4 793 84
AB084153.1 - 2 - 3 375 84
EU826598.1 - 2 - 3 375 84
EU826599.1 - 2 - 3 375 84
EU826600.1 - 2 - 3 375 84
KP192328.1 - 2 - 3 459 84
KP192331.1 - 2 - 3 459 84
M55614.1 - 2 - 3 387 84
M80634.1 - 3 - 4 793 84
M87770.1 - 3 - 4 648 84
M87771.1 - 3 - 4 862 84
M87772.1 - 3 - 4 650 84
M97193.1 - 3 - 4 986 84
U11814.1 - 3 - 4 633 84
X52832.1 - 3 - 4 554 84
X56191.1 - 2 - 3 399 84
Z71929.1 - 2 - 3 387 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000012050.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10001711.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene FGFR2

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
MAPK cascade
angiogenesis
ureteric bud development
in utero embryonic development
epithelial to mesenchymal transition
positive regulation of mesenchymal cell proliferation
outflow tract septum morphogenesis
membranous septum morphogenesis
apoptotic process
cell-cell signaling
axonogenesis
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
regulation of smoothened signaling pathway
post-embryonic development
embryonic pattern specification
organ morphogenesis
regulation of cell fate commitment
positive regulation of phospholipase activity
regulation of phosphatidylinositol 3-kinase signaling
morphogenesis of embryonic epithelium
peptidyl-tyrosine phosphorylation
orbitofrontal cortex development
ventricular zone neuroblast division
pyramidal neuron development
gland morphogenesis
positive regulation of Wnt signaling pathway
bone mineralization
lung development
epithelial cell differentiation
midbrain development
otic vesicle formation
hair follicle morphogenesis
lacrimal gland development
regulation of osteoblast proliferation
multicellular organism growth
organ growth
fibroblast growth factor receptor signaling pathway involved in negative regulation of apoptotic process in bone marrow
fibroblast growth factor receptor signaling pathway involved in hemopoiesis
fibroblast growth factor receptor signaling pathway involved in positive regulation of cell proliferation in bone marrow
fibroblast growth factor receptor signaling pathway involved in orbitofrontal cortex development
phosphatidylinositol-3-phosphate biosynthetic process
regulation of multicellular organism growth
regulation of fibroblast growth factor receptor signaling pathway
inner ear morphogenesis
odontogenesis
positive regulation of MAPK cascade
positive regulation of GTPase activity
cell fate commitment
regulation of osteoblast differentiation
positive regulation of cell cycle
positive regulation of transcription from RNA polymerase II promoter
protein autophosphorylation
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
lung alveolus development
mesodermal cell differentiation
embryonic digestive tract morphogenesis
embryonic organ morphogenesis
digestive tract development
embryonic organ development
reproductive structure development
positive regulation of smooth muscle cell proliferation
embryonic cranial skeleton morphogenesis
skeletal system morphogenesis
epidermis morphogenesis
branching morphogenesis of a nerve
mesenchymal cell differentiation
positive regulation of epithelial cell proliferation
negative regulation of epithelial cell proliferation
regulation of smooth muscle cell differentiation
positive regulation of cell division
ventricular cardiac muscle tissue morphogenesis
positive regulation of cardiac muscle cell proliferation
limb bud formation
bone development
bone morphogenesis
branching involved in prostate gland morphogenesis
branching involved in salivary gland morphogenesis
bud elongation involved in lung branching
lung lobe morphogenesis
lung-associated mesenchyme development
positive regulation of epithelial cell proliferation involved in lung morphogenesis
prostate gland morphogenesis
prostate epithelial cord elongation
prostate epithelial cord arborization involved in prostate glandular acinus morphogenesis
squamous basal epithelial stem cell differentiation involved in prostate gland acinus development
fibroblast growth factor receptor signaling pathway involved in mammary gland specification
lateral sprouting from an epithelium
mammary gland bud formation
epithelial cell proliferation involved in salivary gland morphogenesis
branch elongation involved in salivary gland morphogenesis
branching involved in labyrinthine layer morphogenesis
regulation of branching involved in prostate gland morphogenesis
regulation of morphogenesis of a branching structure
mesenchymal cell differentiation involved in lung development
mesenchymal cell proliferation involved in lung development
regulation of ERK1 and ERK2 cascade
positive regulation of ERK1 and ERK2 cascade
positive regulation of canonical Wnt signaling pathway
protein tyrosine kinase activity
fibroblast growth factor-activated receptor activity
Ras guanyl-nucleotide exchange factor activity
protein binding
ATP binding
heparin binding
1-phosphatidylinositol-3-kinase activity
fibroblast growth factor binding
protein homodimerization activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR10022624.hg.1 ENST00000346997.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10022624.hg.1 ENST00000351936.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10022624.hg.1 ENST00000357555.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10022624.hg.1 ENST00000358487.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10022624.hg.1 ENST00000359354.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10022624.hg.1 ENST00000360144.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10022624.hg.1 ENST00000369056.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10022624.hg.1 ENST00000369060.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10022624.hg.1 ENST00000369061.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10022624.hg.1 ENST00000369062.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10022624.hg.1 ENST00000457416.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10022624.hg.1 NM_000141.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10022624.hg.1 NM_001144913.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10022624.hg.1 NM_001144914.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10022624.hg.1 NM_001144915.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10022624.hg.1 NM_001144917.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10022624.hg.1 NM_001144919.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10022624.hg.1 NM_022970.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10022624.hg.1 TC10001711.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000336553.10 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000346997.6 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000351936.10 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000357555.9 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000358487.9 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000359354.6 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000360144.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000369056.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000369058.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000369060.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000369061.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000457416.6 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000490349.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000491475.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000611527.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000613048.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000613324.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 NM_000141.4 Refseq
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 NM_001144913.1 Refseq
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 NM_001144914.1 Refseq
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 NM_001144915.1 Refseq
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 NM_001144917.1 Refseq
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 NM_001144919.1 Refseq
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 NM_022970.3 Refseq
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 NM_023029.2 Refseq
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000336553.10 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000346997.6 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000351936.10 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000357555.9 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000358487.9 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000359354.6 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000360144.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000369056.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000369058.7 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000369060.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000369061.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000457416.6 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000490349.5 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000491475.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000611527.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000613048.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 ENST00000613324.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 NM_000141.4 Refseq
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 NM_001144913.1 Refseq
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 NM_001144914.1 Refseq
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 NM_001144915.1 Refseq
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 NM_001144917.1 Refseq
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 NM_001144919.1 Refseq
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 NM_022970.3 Refseq
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 NM_023029.2 Refseq
Affymetrix Human Clariom D Assay PSR1000182431.hg.1 TC1000012050.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline