Assay Details
| Gene Symbol: MED12L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053002.5 | NP_443728.3 | 9 - 10 | 1480 | 69 | |
| XM_006713487.3 | XP_006713550.1 | 10 - 11 | 1752 | 69 | ||
| XM_011512390.2 | XP_011510692.1 | 10 - 11 | 1752 | 69 | ||
| XM_011512394.2 | XP_011510696.1 | 10 - 11 | 1752 | 69 | ||
| XM_011512399.2 | XP_011510701.1 | 10 - 11 | 1752 | 69 | ||
| XM_017005676.1 | XP_016861165.1 | 10 - 11 | 1752 | 69 | ||
| XM_017005677.1 | XP_016861166.1 | 10 - 11 | 1752 | 69 | ||
| XM_017005678.1 | XP_016861167.1 | 10 - 11 | 1752 | 69 | ||
| XM_017005679.1 | XP_016861168.1 | 9 - 10 | 1482 | 69 | ||
| XM_017005680.1 | XP_016861169.1 | 10 - 11 | 1752 | 69 | ||
| GenBank mRNA | AF388364.1 | - | 9 - 10 | 1884 | 69 | |
| AF388365.1 | - | 9 - 10 | 1884 | 69 | ||
| AF399708.1 | - | 9 - 10 | 1389 | 69 | ||
| BC144559.1 | - | 7 - 8 | 964 | 9053087 | 69 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000822.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000273432.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000309237.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000363124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000422248.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000468305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000468305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000468305.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000469768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000469768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000469768.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000474524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000480026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000480026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000488092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000488092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000491549.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000491549.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000491549.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000491549.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000491549.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | ENST00000491549.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009179.hg.1 | NM_053002.5 | Refseq |

MED12L
Genomic Map