This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01553794_g1
See other GTF2F2 GE Assays ›
Gene Symbol:
GTF2F2
Gene Name:
general transcription factor IIF subunit 2
Gene Aliases:
BTF4, RAP30, TF2F2, TFIIF
Chromosome Location:
Chr.13: 45119830 - 45284105 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01553794_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GTF2F2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_004128.2 NP_004119.1 7 - 8 114
XM_011535052.2 XP_011533354.1 8 - 9 114
XM_017020551.1 XP_016876040.1 9 - 10 114
GenBank mRNA AK291545.1 - 7 - 8 114
BC001771.1 - 7 - 9 750 3351446 114
BT019525.1 - 7 - 8 114
X16901.1 - 7 - 8 748 114
X59745.1 - 7 - 9 629 114

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300007052.hg.1
This assay targets the same location as HTA transcript cluster(s) TC13000171.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GTF2F2

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
7-methylguanosine mRNA capping
fibroblast growth factor receptor signaling pathway
gene expression
positive regulation of transcription elongation from RNA polymerase II promoter
snRNA transcription from RNA polymerase II promoter
positive regulation of RNA polymerase II transcriptional preinitiation complex assembly
positive regulation of viral transcription
positive regulation of transcription initiation from RNA polymerase II promoter
transcription factor activity, core RNA polymerase II binding
DNA binding
helicase activity
protein binding
ATP binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000340473.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000340473.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000340473.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000340473.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000340473.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000340473.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000340473.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000340473.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000340473.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000340473.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000340473.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000340473.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000461904.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000461904.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000461904.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000461904.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000461904.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000494087.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000494087.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000494087.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 ENST00000494087.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 NM_004128.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 NM_004128.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 NM_004128.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 NM_004128.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 NM_004128.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 NM_004128.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 NM_004128.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 NM_004128.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 NM_004128.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 NM_004128.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300007052.hg.1 NM_004128.2 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 153454 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0