This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01554177_m1
See other EIF4G3 GE Assays ›
Gene Symbol:
EIF4G3
Gene Name:
eukaryotic translation initiation factor 4 gamma 3
Gene Aliases:
eIF-4G 3, eIF4G 3, eIF4GII
Chromosome Location:
Chr.1: 20806292 - 21177320 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01554177_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EIF4G3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001198801.1 NP_001185730.1 31 - 32 4926 94
NM_001198802.1 NP_001185731.1 31 - 32 4836 94
NM_003760.4 NP_003751.2 28 - 29 4452 94
XM_011542356.2 XP_011540658.1 34 - 35 5526 94
XM_011542361.2 XP_011540663.1 33 - 34 5415 94
XM_011542370.1 XP_011540672.1 34 - 35 5529 94
XM_011542371.1 XP_011540673.1 32 - 33 5271 94
XM_011542374.1 XP_011540676.1 31 - 32 5238 94
XM_011542375.2 XP_011540677.1 31 - 32 6826 94
XM_011542381.2 XP_011540683.1 21 - 22 2912 94
XM_017002672.1 XP_016858161.1 35 - 36 7064 94
XM_017002673.1 XP_016858162.1 33 - 34 5508 94
XM_017002674.1 XP_016858163.1 33 - 34 5505 94
XM_017002675.1 XP_016858164.1 33 - 34 5496 94
XM_017002676.1 XP_016858165.1 32 - 33 5475 94
XM_017002677.1 XP_016858166.1 32 - 33 5352 94
XM_017002678.1 XP_016858167.1 33 - 34 5418 94
XM_017002679.1 XP_016858168.1 33 - 34 5415 94
XM_017002680.1 XP_016858169.1 30 - 31 4445 94
XM_017002681.1 XP_016858170.1 33 - 34 5412 94
XM_017002682.1 XP_016858171.1 32 - 33 5394 94
XM_017002683.1 XP_016858172.1 32 - 33 5394 94
XM_017002684.1 XP_016858173.1 33 - 34 6920 94
XM_017002685.1 XP_016858174.1 32 - 33 5385 94
XM_017002686.1 XP_016858175.1 32 - 33 5382 94
XM_017002687.1 XP_016858176.1 32 - 33 5382 94
XM_017002688.1 XP_016858177.1 29 - 30 4411 94
XM_017002689.1 XP_016858178.1 34 - 35 5529 94
XM_017002690.1 XP_016858179.1 32 - 33 5379 94
XM_017002691.1 XP_016858180.1 31 - 32 5364 94
XM_017002692.1 XP_016858181.1 31 - 32 5361 94
XM_017002693.1 XP_016858182.1 30 - 31 5232 94
XM_017002694.1 XP_016858183.1 32 - 33 5304 94
XM_017002695.1 XP_016858184.1 32 - 33 5301 94
XM_017002696.1 XP_016858185.1 31 - 32 5283 94
XM_017002697.1 XP_016858186.1 31 - 32 5280 94
XM_017002698.1 XP_016858187.1 31 - 32 5271 94
XM_017002699.1 XP_016858188.1 33 - 34 5418 94
XM_017002700.1 XP_016858189.1 30 - 31 5250 94
XM_017002701.1 XP_016858190.1 30 - 31 5247 94
XM_017002702.1 XP_016858191.1 30 - 31 5181 94
XM_017002703.1 XP_016858192.1 28 - 29 4342 94
XM_017002704.1 XP_016858193.1 32 - 33 5394 94
XM_017002705.1 XP_016858194.1 33 - 34 4689 94
XM_017002706.1 XP_016858195.1 32 - 33 4578 94
XM_017002707.1 XP_016858196.1 30 - 31 4521 94
XM_017002708.1 XP_016858197.1 29 - 30 4410 94
XM_017002709.1 XP_016858198.1 31 - 32 5273 94
GenBank mRNA AB209119.1 - 31 - 32 4795 94
AF012072.2 - 28 - 29 4452 94
AK294883.1 - 22 - 23 3699 94
AK302873.1 - 19 - 20 2693 94
BC094683.1 - 26 - 27 3459 6188273 94
BC136643.1 - 28 - 29 4375 9020633 94
Z34918.1 - 12 - 13 1538 94

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100013194.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01002320.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EIF4G3

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

translational initiation
regulation of translational initiation
viral process
RNA cap binding
translation initiation factor activity
translation factor activity, RNA binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01035583.hg.1 TC01002320.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01035583.hg.1 ENST00000264211.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01035583.hg.1 ENST00000374935.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01035583.hg.1 ENST00000374937.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01035583.hg.1 ENST00000400415.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01035583.hg.1 ENST00000400422.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01035583.hg.1 ENST00000536266.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01035583.hg.1 ENST00000537738.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01035583.hg.1 NM_001198801.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01035583.hg.1 NM_001198802.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01035583.hg.1 NM_003760.4 Refseq
Affymetrix Human Clariom D Assay PSR0100197721.hg.1 ENST00000264211.12 Ensembl
Affymetrix Human Clariom D Assay PSR0100197721.hg.1 ENST00000374935.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100197721.hg.1 ENST00000400422.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100197721.hg.1 ENST00000602326.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100197721.hg.1 NM_001198801.1 Refseq
Affymetrix Human Clariom D Assay PSR0100197721.hg.1 NM_001198802.1 Refseq
Affymetrix Human Clariom D Assay PSR0100197721.hg.1 NM_003760.4 Refseq
Affymetrix Human Clariom D Assay PSR0100197721.hg.1 ENST00000264211.12 Ensembl
Affymetrix Human Clariom D Assay PSR0100197721.hg.1 ENST00000374935.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100197721.hg.1 ENST00000400422.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100197721.hg.1 ENST00000602326.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100197721.hg.1 NM_001198801.1 Refseq
Affymetrix Human Clariom D Assay PSR0100197721.hg.1 NM_001198802.1 Refseq
Affymetrix Human Clariom D Assay PSR0100197721.hg.1 NM_003760.4 Refseq
Affymetrix Human Clariom D Assay PSR0100197721.hg.1 TC0100013194.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0