This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01554360_m1
See other SDS GE Assays ›
Gene Symbol:
SDS
Gene Name:
serine dehydratase
Gene Aliases:
SDH
Chromosome Location:
Chr.12: 113392445 - 113403887 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01554360_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SDS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_006843.2 NP_006834.2 1 - 2 123 95
GenBank mRNA AK292760.1 - 1 - 2 119 95
BC020750.1 - 1 - 2 122 4662951 95
BG534053.1 - 1 - 2 123 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200011994.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12001989.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK292760.1,BC020750.1,NM_006843.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SDS

Panther Classification:

Molecular Function -

dehydratase

Gene Ontology Categories:

Function(s) Process(es)

gluconeogenesis
L-serine catabolic process
pyruvate biosynthetic process
L-serine ammonia-lyase activity
L-threonine ammonia-lyase activity
pyridoxal phosphate binding
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR12026140.hg.1 ENST00000257549.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12026140.hg.1 ENST00000446302.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12026140.hg.1 ENST00000547342.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12026140.hg.1 NM_006843.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12026140.hg.1 TC12001989.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR12026141.hg.1 ENST00000257549.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12026141.hg.1 ENST00000446302.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12026141.hg.1 NM_006843.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12026141.hg.1 TC12001989.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000257549.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000257549.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000257549.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000257549.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000257549.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000257549.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000257549.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000257549.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000257549.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000257549.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000546639.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000546639.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000546639.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000546639.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000546785.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000546785.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000546785.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000547342.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000547342.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000547342.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000547342.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000547342.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000547342.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000552280.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000552280.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000552280.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000553112.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000553112.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000553112.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000553112.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000553112.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000553112.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000553112.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000553112.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 ENST00000553112.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 NM_006843.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 NM_006843.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 NM_006843.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 NM_006843.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 NM_006843.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 NM_006843.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 NM_006843.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 NM_006843.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 NM_006843.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011994.hg.1 NM_006843.2 Refseq
Affymetrix Human Clariom D Assay PSR1200193445.hg.1 ENST00000257549.8 Ensembl
Affymetrix Human Clariom D Assay PSR1200193445.hg.1 ENST00000546639.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200193445.hg.1 ENST00000546785.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200193445.hg.1 ENST00000552280.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200193445.hg.1 ENST00000553112.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200193445.hg.1 NM_006843.2 Refseq
Affymetrix Human Clariom D Assay PSR1200193445.hg.1 ENST00000257549.8 Ensembl
Affymetrix Human Clariom D Assay PSR1200193445.hg.1 ENST00000546639.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200193445.hg.1 ENST00000546785.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200193445.hg.1 ENST00000552280.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200193445.hg.1 ENST00000553112.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200193445.hg.1 NM_006843.2 Refseq
Affymetrix Human Clariom D Assay PSR1200193445.hg.1 TC1200011994.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200193443.hg.1 ENST00000257549.8 Ensembl
Affymetrix Human Clariom D Assay PSR1200193443.hg.1 ENST00000546639.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200193443.hg.1 ENST00000546785.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200193443.hg.1 ENST00000547342.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200193443.hg.1 ENST00000552280.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200193443.hg.1 ENST00000553112.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200193443.hg.1 NM_006843.2 Refseq
Affymetrix Human Clariom D Assay PSR1200193443.hg.1 ENST00000257549.8 Ensembl
Affymetrix Human Clariom D Assay PSR1200193443.hg.1 ENST00000546639.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200193443.hg.1 ENST00000546785.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200193443.hg.1 ENST00000547342.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200193443.hg.1 ENST00000552280.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200193443.hg.1 ENST00000553112.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200193443.hg.1 NM_006843.2 Refseq
Affymetrix Human Clariom D Assay PSR1200193443.hg.1 TC1200011994.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000257549.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000257549.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000257549.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000257549.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000257549.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000257549.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000257549.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000257549.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000257549.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000257549.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000546639.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000546639.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000546639.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000546639.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000546785.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000546785.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000546785.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000547342.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000547342.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000547342.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000547342.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000547342.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000547342.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000552280.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000552280.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000552280.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000553112.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000553112.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000553112.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000553112.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000553112.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000553112.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000553112.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000553112.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 ENST00000553112.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011994.hg.1 NM_006843.2 Refseq
Affymetrix Human Clariom D Assay TC1200011994.hg.1 NM_006843.2 Refseq
Affymetrix Human Clariom D Assay TC1200011994.hg.1 NM_006843.2 Refseq
Affymetrix Human Clariom D Assay TC1200011994.hg.1 NM_006843.2 Refseq
Affymetrix Human Clariom D Assay TC1200011994.hg.1 NM_006843.2 Refseq
Affymetrix Human Clariom D Assay TC1200011994.hg.1 NM_006843.2 Refseq
Affymetrix Human Clariom D Assay TC1200011994.hg.1 NM_006843.2 Refseq
Affymetrix Human Clariom D Assay TC1200011994.hg.1 NM_006843.2 Refseq
Affymetrix Human Clariom D Assay TC1200011994.hg.1 NM_006843.2 Refseq
Affymetrix Human Clariom D Assay TC1200011994.hg.1 NM_006843.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0