This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01555493_m1
See other CTH GE Assays ›
Gene Symbol:
CTH
Gene Name:
cystathionine gamma-lyase
Gene Aliases:
-
Chromosome Location:
Chr.1: 70411218 - 70441949 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01555493_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CTH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_153742.4 NP_714964.2 4 - 5 657 154
GenBank mRNA S52028.1 - 4 - 5 492 154

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100008697.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01000757.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CTH

Panther Classification:

Molecular Function -

lyase

Gene Ontology Categories:

Function(s) Process(es)

sulfur amino acid catabolic process
cysteine metabolic process
protein-pyridoxal-5-phosphate linkage via peptidyl-N6-pyridoxal phosphate-L-lysine
cysteine biosynthetic process via cystathionine
cysteine biosynthetic process
transsulfuration
endoplasmic reticulum unfolded protein response
positive regulation of I-kappaB kinase/NF-kappaB signaling
protein sulfhydration
positive regulation of NF-kappaB transcription factor activity
protein homotetramerization
hydrogen sulfide biosynthetic process
'de novo' L-methionine biosynthetic process
positive regulation of aortic smooth muscle cell differentiation
negative regulation of apoptotic signaling pathway
cystathionine gamma-synthase activity
cystathionine gamma-lyase activity
protein binding
calmodulin binding
carbon-sulfur lyase activity
pyridoxal phosphate binding
L-cystine L-cysteine-lyase (deaminating)
homocysteine desulfhydrase activity
L-cysteine desulfhydrase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01012132.hg.1 ENST00000346806.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01012132.hg.1 ENST00000370938.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01012132.hg.1 ENST00000411986.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01012132.hg.1 ENST00000464926.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01012132.hg.1 NM_001190463.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01012132.hg.1 NM_001902.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01012132.hg.1 NM_153742.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01012132.hg.1 TC01000757.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0100161916.hg.1 ENST00000346806.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100161916.hg.1 ENST00000370938.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100161916.hg.1 ENST00000411986.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100161916.hg.1 ENST00000464926.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100161916.hg.1 NM_001190463.1 Refseq
Affymetrix Human Clariom D Assay PSR0100161916.hg.1 NM_001902.5 Refseq
Affymetrix Human Clariom D Assay PSR0100161916.hg.1 NM_153742.4 Refseq
Affymetrix Human Clariom D Assay PSR0100161916.hg.1 ENST00000346806.2 Ensembl
Affymetrix Human Clariom D Assay PSR0100161916.hg.1 ENST00000370938.7 Ensembl
Affymetrix Human Clariom D Assay PSR0100161916.hg.1 ENST00000411986.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100161916.hg.1 ENST00000464926.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100161916.hg.1 NM_001190463.1 Refseq
Affymetrix Human Clariom D Assay PSR0100161916.hg.1 NM_001902.5 Refseq
Affymetrix Human Clariom D Assay PSR0100161916.hg.1 NM_153742.4 Refseq
Affymetrix Human Clariom D Assay PSR0100161916.hg.1 TC0100008697.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline