This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01557334_m1
See other MBOAT1 GE Assays ›
Gene Symbol:
MBOAT1
Gene Name:
membrane bound O-acyltransferase domain containing 1
Gene Aliases:
LPEAT1, LPLAT, LPLAT 1, LPSAT, OACT1, dJ434O11.1
Chromosome Location:
Chr.6: 20099684 - 20212464 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01557334_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MBOAT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080480.2 NP_001073949.1 9 - 10 1247 90
NR_073465.1 6 - 7 967 90
XM_006714999.2 XP_006715062.1 9 - 10 984 90
GenBank mRNA AB305043.1 - 9 - 10 1017 90
AK093994.1 - 3 - 4 446 90
AK131269.1 - 9 - 10 1210 90
AK296857.1 - 6 - 7 967 90
BC150650.1 - 9 - 10 1193 9057021 90
BC150652.1 - 9 - 10 1193 9057023 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600010991.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06001313.hg.1

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

acetyltransferase

Gene Ontology Categories:

Function(s) Process(es)

phospholipid biosynthetic process
phosphatidylserine acyl-chain remodeling
phosphatidylethanolamine acyl-chain remodeling
1-acylglycerol-3-phosphate O-acyltransferase activity
2-acylglycerol-3-phosphate O-acyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06015724.hg.1 TC06001313.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR06015724.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06015724.hg.1 ENST00000541730.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06015724.hg.1 NM_001080480.2 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Clariom D Assay TC0600010991.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Clariom D Assay TC0600010991.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Clariom D Assay TC0600010991.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Clariom D Assay TC0600010991.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Clariom D Assay TC0600010991.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Clariom D Assay TC0600010991.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Clariom D Assay TC0600010991.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Clariom D Assay TC0600010991.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Clariom D Assay TC0600010991.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Clariom D Assay TC0600010991.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Clariom D Assay TC0600010991.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Clariom D Assay TC0600010991.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Clariom D Assay TC0600010991.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Clariom D Assay TC0600010991.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NM_001080480.2 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Clariom D Assay TC0600010991.hg.1 NR_073465.1 Refseq
Affymetrix Human Clariom D Assay PSR0600174547.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Clariom D Assay PSR0600174547.hg.1 NM_001080480.2 Refseq
Affymetrix Human Clariom D Assay PSR0600174547.hg.1 NR_073465.1 Refseq
Affymetrix Human Clariom D Assay PSR0600174547.hg.1 ENST00000324607.7 Ensembl
Affymetrix Human Clariom D Assay PSR0600174547.hg.1 NM_001080480.2 Refseq
Affymetrix Human Clariom D Assay PSR0600174547.hg.1 NR_073465.1 Refseq
Affymetrix Human Clariom D Assay PSR0600174547.hg.1 TC0600010991.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0