This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01557502_g1
See other ARHGAP36 GE Assays ›
Gene Symbol:
ARHGAP36
Gene Name:
Rho GTPase activating protein 36
Gene Aliases:
-
Chromosome Location:
Chr.X: 131058242 - 131089885 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01557502_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARHGAP36
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001282607.1 NP_001269536.1 4 - 5 665 65
NM_144967.3 NP_659404.2 4 - 5 901 65
XM_006724729.1 XP_006724792.1 3 - 4 372 65
XM_011531280.1 XP_011529582.1 3 - 4 389 65
GenBank mRNA AK054620.1 - 1 - 2 261 65
AK128203.1 - 4 - 5 901 65
AK294274.1 - 4 - 5 624 65
AK296748.1 - 4 - 5 643 65
BC063790.1 - 4 - 5 500 6196091 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0X00008415.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X000625.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ARHGAP36

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
GTPase activator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000370921.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000370921.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000370921.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000370921.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000370921.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000370921.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000370921.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000370921.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000370921.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000370921.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000370922.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000370922.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000370922.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000370922.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000370922.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000370922.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000370922.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000370922.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000370922.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000370922.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000370922.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000370922.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000370922.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000370922.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000412432.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000412432.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000412432.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000412432.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000412432.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000412432.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000412432.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000412432.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000412432.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000412432.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000412432.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000412432.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000412432.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000423277.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000423277.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000423277.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000423277.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 ENST00000423277.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_001282607.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_001282607.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_001282607.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_001282607.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_001282607.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_001282607.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_001282607.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_001282607.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_001282607.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_001282607.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_001282607.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_001282607.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_001282607.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_001282607.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_001282607.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X008277.hg.1 TC0X000625.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR0X008277.hg.1 ENST00000276211.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X008277.hg.1 ENST00000370921.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X008277.hg.1 ENST00000370922.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X008277.hg.1 ENST00000412432.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X008277.hg.1 NM_144967.3 Refseq
Affymetrix Human Clariom D Assay PSR0X00157330.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Clariom D Assay PSR0X00157330.hg.1 ENST00000370921.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00157330.hg.1 ENST00000370922.5 Ensembl
Affymetrix Human Clariom D Assay PSR0X00157330.hg.1 ENST00000412432.6 Ensembl
Affymetrix Human Clariom D Assay PSR0X00157330.hg.1 ENST00000423277.5 Ensembl
Affymetrix Human Clariom D Assay PSR0X00157330.hg.1 NM_001282607.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00157330.hg.1 NM_144967.3 Refseq
Affymetrix Human Clariom D Assay PSR0X00157330.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Clariom D Assay PSR0X00157330.hg.1 ENST00000370921.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00157330.hg.1 ENST00000370922.5 Ensembl
Affymetrix Human Clariom D Assay PSR0X00157330.hg.1 ENST00000412432.6 Ensembl
Affymetrix Human Clariom D Assay PSR0X00157330.hg.1 ENST00000423277.5 Ensembl
Affymetrix Human Clariom D Assay PSR0X00157330.hg.1 NM_001282607.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00157330.hg.1 NM_144967.3 Refseq
Affymetrix Human Clariom D Assay PSR0X00157330.hg.1 TC0X00008415.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000276211.9 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000370921.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000370921.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000370921.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000370921.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000370921.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000370921.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000370921.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000370921.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000370921.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000370921.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000370922.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000370922.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000370922.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000370922.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000370922.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000370922.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000370922.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000370922.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000370922.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000370922.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000370922.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000370922.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000370922.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000370922.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000412432.6 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000412432.6 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000412432.6 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000412432.6 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000412432.6 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000412432.6 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000412432.6 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000412432.6 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000412432.6 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000412432.6 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000412432.6 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000412432.6 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000412432.6 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000423277.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000423277.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000423277.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000423277.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 ENST00000423277.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_001282607.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_001282607.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_001282607.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_001282607.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_001282607.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_001282607.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_001282607.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_001282607.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_001282607.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_001282607.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_001282607.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_001282607.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_001282607.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_001282607.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_001282607.1 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_144967.3 Refseq
Affymetrix Human Clariom D Assay TC0X00008415.hg.1 NM_144967.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0