This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01558716_g1
See other LMF2 GE Assays ›
Gene Symbol:
LMF2
Gene Name:
lipase maturation factor 2
Gene Aliases:
TMEM112B, TMEM153
Chromosome Location:
Chr.22: 50502947 - 50507840 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01558716_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LMF2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_033200.2 NP_149977.2 5 - 6 806 90
XM_005261955.3 XP_005262012.1 5 - 6 888 90
XM_006724426.3 XP_006724489.1 4 - 5 1252 90
XM_006724427.3 XP_006724490.1 5 - 6 803 90
XM_017029077.1 XP_016884566.1 5 - 6 940 90
GenBank mRNA BC002942.2 - 5 - 6 780 3954961 90
BC014652.1 - 5 - 6 869 4562676 90
BC021143.1 - 5 - 6 780 4872642 90
CR456605.1 - 5 - 6 790 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2200009161.hg.1
This assay targets the same location as HTA transcript cluster(s) TC22000914.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene LMF2

Gene Ontology Categories:

Process(es)

protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR22015597.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22015597.hg.1 ENST00000380796.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22015597.hg.1 ENST00000474879.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22015597.hg.1 ENST00000505981.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22015597.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22015597.hg.1 TC22000914.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000505981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000505981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000505981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000505981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000505981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000505981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000507607.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000507607.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000507607.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000507607.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000507607.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000504717.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000505981.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000505981.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000505981.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000505981.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000505981.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000505981.5 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000507607.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000507607.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000507607.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000507607.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000507607.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay TC2200009161.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay PSR2200166503.hg.1 TC2200009161.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2200166503.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200166503.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay PSR2200166503.hg.1 ENST00000505981.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200166503.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Clariom D Assay PSR2200166503.hg.1 NM_033200.2 Refseq
Affymetrix Human Clariom D Assay PSR2200166503.hg.1 ENST00000216080.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200166503.hg.1 ENST00000474879.6 Ensembl
Affymetrix Human Clariom D Assay PSR2200166503.hg.1 ENST00000505981.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200166503.hg.1 ENST00000514938.1 Ensembl
Affymetrix Human Clariom D Assay PSR2200166503.hg.1 NM_033200.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0