This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01560740_m1
See other CCT6P3 GE Assays ›
Gene Symbol:
CCT6P3
Gene Name:
chaperonin containing TCP1 subunit 6 pseudogene 3
Gene Aliases:
CCT6-3P
Chromosome Location:
Chr.7: 65038354 - 65074713 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01560740_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CCT6P3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_033416.1 1 - 2 215 70
GenBank mRNA BX649060.1 - 1 - 2 216 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700007795.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07000388.hg.1

Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Homo sapiens 1700 Probe spans exons

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000275603.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000275603.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000275603.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000275603.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000275603.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000275603.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000275603.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000275603.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000275603.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000275603.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000275603.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000275603.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000275603.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000275603.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000275603.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000275603.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000275603.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000275603.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000275603.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000275603.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000275603.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000275603.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000275603.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000335503.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000335503.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000335503.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000335503.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000335503.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000335503.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000335503.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000335503.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000335503.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000335503.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000335503.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000335503.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000335503.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000335503.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000335503.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000335503.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000335503.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000335503.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000335503.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000335503.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000462133.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000462133.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000462133.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000462133.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000462133.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000462133.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000462133.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000466479.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000466479.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000466479.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000466479.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000466479.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000466479.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000466572.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000466572.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000466572.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000482776.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000482776.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000482776.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000482776.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000482776.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000492618.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000492618.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000492618.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000492618.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000492618.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000493855.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000493855.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000493855.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000494736.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000494736.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000494736.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000494736.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 ENST00000494736.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001009186.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001009186.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001009186.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001009186.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001009186.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001009186.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001009186.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001009186.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001009186.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001009186.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001009186.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001009186.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001009186.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001009186.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001009186.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001009186.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001009186.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001009186.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001009186.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001009186.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001762.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001762.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001762.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001762.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001762.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001762.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001762.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001762.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001762.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001762.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001762.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001762.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001762.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001762.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001762.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001762.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001762.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001762.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001762.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001762.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001762.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007630.hg.1 NM_001762.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000419314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000419314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000419314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000419314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000419314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000419314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000419314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000419314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000419314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000419314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000419314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000419314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000419314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000419314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000419314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000419314.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000426828.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000426828.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000426828.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000426828.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000426828.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000426828.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000426828.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000426828.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000426828.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000426828.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000426828.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000426828.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000426828.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000426828.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000430369.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000452026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000452026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000452026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000452026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000452026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000452026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000452026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000452026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000452026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000452026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000452026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000452026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000452026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 ENST00000452026.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 NR_033416.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 NR_033416.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 NR_033416.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 NR_033416.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 NR_033416.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 NR_033416.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 NR_033416.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 NR_033416.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 NR_033416.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 NR_033416.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 NR_033416.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 NR_033416.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 NR_033416.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700007795.hg.1 NR_033416.1 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 236043 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline