This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01560900_m1
See other TNFRSF21 GE Assays ›
Gene Symbol:
TNFRSF21
Gene Name:
TNF receptor superfamily member 21
Gene Aliases:
BM-018, CD358, DR6
Chromosome Location:
Chr.6: 47231527 - 47309976 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01560900_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TNFRSF21
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_014452.4 NP_055267.1 4 - 5 1947 129
GenBank mRNA AB209394.1 - 3 - 4 3138 129
AF068868.1 - 4 - 5 1511 129
AJ420531.1 - 3 - 4 1316 129
AK001504.1 - 4 - 5 1905 129
AK315560.1 - 4 - 5 1911 129
AY358304.1 - 4 - 5 1835 129
BC010241.1 - 3 - 4 1104 4154494 129
BC017730.2 - 4 - 5 1522 3847246 129
BC021572.1 - 4 - 5 1823 4874397 129
BT007420.1 - 4 - 5 1511 129

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600011960.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06001777.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TNFRSF21

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

B cell apoptotic process
adaptive immune response
apoptotic process
inflammatory response
immune response
humoral immune response
negative regulation of B cell proliferation
negative regulation of myelination
response to lipopolysaccharide
tumor necrosis factor-mediated signaling pathway
regulation of cell proliferation
negative regulation of T cell proliferation
myelination
regulation of apoptotic process
regulation of oligodendrocyte differentiation
T cell receptor signaling pathway
neuron apoptotic process
cellular response to tumor necrosis factor
apoptotic signaling pathway
oligodendrocyte apoptotic process
negative regulation of interleukin-5 secretion
negative regulation of interleukin-13 secretion
negative regulation of interleukin-10 secretion
tumor necrosis factor-activated receptor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600011960.hg.1 ENST00000296861.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011960.hg.1 ENST00000296861.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011960.hg.1 ENST00000296861.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011960.hg.1 ENST00000296861.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011960.hg.1 ENST00000296861.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011960.hg.1 ENST00000296861.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011960.hg.1 ENST00000296861.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011960.hg.1 ENST00000296861.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011960.hg.1 ENST00000296861.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011960.hg.1 ENST00000296861.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600011960.hg.1 NM_014452.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011960.hg.1 NM_014452.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011960.hg.1 NM_014452.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011960.hg.1 NM_014452.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011960.hg.1 NM_014452.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011960.hg.1 NM_014452.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011960.hg.1 NM_014452.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011960.hg.1 NM_014452.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011960.hg.1 NM_014452.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011960.hg.1 NM_014452.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600011960.hg.1 NM_014452.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-bs78j:80/100.66.77.131:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0