This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01561210_g1
See other TAP2 GE Assays ›
Gene Symbol:
TAP2
Gene Name:
transporter 2, ATP binding cassette subfamily B member
Gene Aliases:
ABC18, ABCB3, APT2, D6S217E, PSF-2, PSF2, RING11
Chromosome Location:
Chr.6: 32821833 - 32838823 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01561210_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TAP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000544.3 NP_000535.3 2 - 3 617 66
NM_001290043.1 NP_001276972.1 2 - 3 670 66
NM_018833.2 NP_061313.2 2 - 3 617 66
GenBank mRNA AB073779.1 - 1 - 2 495 66
AB208953.1 - 2 - 3 564 66
AF078671.1 - 2 - 3 574 66
AF105151.1 - 2 - 3 574 66
AK222823.1 - 1 - 2 526 66
AK223300.1 - 2 - 3 577 66
AK225657.1 - 2 - 3 598 66
AK225693.1 - 2 - 3 577 66
AK292963.1 - 2 - 3 600 66
BC002751.2 - 1 - 2 503 66
BT009906.1 - 1 - 2 495 66
FM246456.1 - 1 - 2 495 66
M74447.1 - 2 - 3 591 66
U07844.1 - 1 - 2 305 66
Z22935.1 - 2 - 3 523 66
Z22936.1 - 2 - 3 571 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600014275.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06004126.hg.1, TC6_apd_hap1000130.hg.1, TC6_cox_hap2000257.hg.1, TC6_dbb_hap3000242.hg.1, TC6_mann_hap4000165.hg.1, TC6_mcf_hap5000225.hg.1, TC6_qbl_hap6000185.hg.1, TC6_ssto_hap7000213.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TAP2

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter

Gene Ontology Categories:

Function(s) Process(es)

adaptive immune response
antigen processing and presentation of peptide antigen via MHC class I
antigen processing and presentation of exogenous protein antigen via MHC class Ib, TAP-dependent
antigen processing and presentation of endogenous peptide antigen via MHC class I via ER pathway, TAP-dependent
antigen processing and presentation of endogenous peptide antigen via MHC class Ib via ER pathway, TAP-dependent
positive regulation of antigen processing and presentation of peptide antigen via MHC class I
intracellular transport of viral protein in host cell
antigen processing and presentation of endogenous peptide antigen via MHC class I
cytosol to ER transport
peptide antigen transport
transmembrane transport
transporter activity
protein binding
ATP binding
peptide transporter activity
peptide antigen-transporting ATPase activity
MHC class Ib protein binding
TAP1 binding
tapasin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR6_apd_hap1001719.hg.1 NM_000544.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_apd_hap1001719.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_apd_hap1001719.hg.1 TC6_apd_hap1000130.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_ssto_hap7003485.hg.1 NM_000544.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_ssto_hap7003485.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_ssto_hap7003485.hg.1 TC6_ssto_hap7000213.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6003726.hg.1 NM_000544.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6003726.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6003726.hg.1 TC6_qbl_hap6000185.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000464100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000464100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000464100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000464100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000464100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000464100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000485701.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000485701.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000485701.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06018767.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06018767.hg.1 ENST00000374899.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06018767.hg.1 NM_000544.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06018767.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06018767.hg.1 TC06004126.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_mann_hap4003015.hg.1 NM_000544.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_mann_hap4003015.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_mann_hap4003015.hg.1 TC6_mann_hap4000165.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_mcf_hap5003551.hg.1 NM_000544.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_mcf_hap5003551.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_mcf_hap5003551.hg.1 TC6_mcf_hap5000225.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2003911.hg.1 NM_000544.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2003911.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2003911.hg.1 TC6_cox_hap2000257.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_dbb_hap3003664.hg.1 NM_000544.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_dbb_hap3003664.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_dbb_hap3003664.hg.1 TC6_dbb_hap3000242.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000464100.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000464100.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000464100.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000464100.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000464100.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000464100.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000485701.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000485701.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000485701.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay PSR0600201520.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay PSR0600201520.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600201520.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600201520.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay PSR0600201520.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay PSR0600201520.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600201520.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600201520.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay PSR0600201520.hg.1 TC0600014275.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline