This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01561213_m1
See other TAP2 GE Assays ›
Gene Symbol:
TAP2
Gene Name:
transporter 2, ATP binding cassette subfamily B member
Gene Aliases:
ABC18, ABCB3, APT2, D6S217E, PSF-2, PSF2, RING11
Chromosome Location:
Chr.6: 32821833 - 32838823 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01561213_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TAP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000544.3 NP_000535.3 5 - 6 1066 64
NM_001290043.1 NP_001276972.1 5 - 6 1119 64
NM_018833.2 NP_061313.2 5 - 6 1066 64
GenBank mRNA AB073779.1 - 4 - 5 944 64
AB208953.1 - 5 - 6 1013 64
AF078671.1 - 5 - 6 1023 64
AF105151.1 - 5 - 6 1023 64
AK222823.1 - 4 - 5 975 64
AK223300.1 - 5 - 6 1026 64
AK225657.1 - 5 - 6 1047 64
AK225693.1 - 5 - 6 1026 64
AK292963.1 - 5 - 6 1049 64
BC002751.2 - 4 - 5 952 64
BT009906.1 - 4 - 5 944 64
FM246456.1 - 4 - 5 944 64
M74447.1 - 5 - 6 1040 64
M84748.1 - 5 - 6 972 64
U07844.1 - 4 - 5 754 64
Z22935.1 - 5 - 6 972 64
Z22936.1 - 5 - 6 1020 64

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TAP2

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter

Gene Ontology Categories:

Function(s) Process(es)

adaptive immune response
antigen processing and presentation of peptide antigen via MHC class I
antigen processing and presentation of exogenous protein antigen via MHC class Ib, TAP-dependent
antigen processing and presentation of endogenous peptide antigen via MHC class I via ER pathway, TAP-dependent
antigen processing and presentation of endogenous peptide antigen via MHC class Ib via ER pathway, TAP-dependent
positive regulation of antigen processing and presentation of peptide antigen via MHC class I
intracellular transport of viral protein in host cell
antigen processing and presentation of endogenous peptide antigen via MHC class I
cytosol to ER transport
peptide antigen transport
transmembrane transport
transporter activity
protein binding
ATP binding
peptide transporter activity
peptide antigen-transporting ATPase activity
MHC class Ib protein binding
TAP1 binding
tapasin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000464100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000464100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000464100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000464100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000464100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000464100.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000485701.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000485701.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000485701.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374897.2 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000374899.8 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000464100.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000464100.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000464100.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000464100.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000464100.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000464100.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000485701.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000485701.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000485701.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 ENST00000620123.4 Ensembl
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq
Affymetrix Human Clariom D Assay TC0600014275.hg.1 NM_018833.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline