This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01562316_m1
See other SPIB GE Assays ›
Gene Symbol:
SPIB
Gene Name:
Spi-B transcription factor
Gene Aliases:
SPI-B
Chromosome Location:
Chr.19: 50418938 - 50431053 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01562316_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SPIB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001243999.1 NP_001230928.1 5 - 6 507 81
GenBank mRNA X96998.1 - 5 - 6 487 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

winged helix/forkhead transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
cell differentiation
RNA polymerase II transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000270632.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000270632.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000270632.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000270632.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000270632.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000270632.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000270632.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000270632.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000270632.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000270632.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000270632.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000439922.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000439922.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000439922.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000439922.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000439922.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000439922.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000439922.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000439922.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000439922.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000439922.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000594188.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000594188.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000594188.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000594188.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000594188.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000594685.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000594685.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000594685.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000594685.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000594685.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000595883.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000595883.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000595883.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000595883.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000595883.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000595883.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000595883.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000595883.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000595883.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000595883.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000595883.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000595883.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000595883.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000596074.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000596074.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000596074.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000596074.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000596074.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000596074.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000596074.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000596074.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000597855.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000597855.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000597855.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000597855.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000597855.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000597855.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000597855.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000599923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000599923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000599923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 ENST00000599923.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001243998.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001243998.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001243998.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001243998.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001243998.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001243998.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001243998.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001243998.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001243998.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001243998.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001243999.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001243999.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001243999.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001243999.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001243999.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001243999.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001243999.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001243999.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001243999.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001243999.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001243999.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001243999.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001244000.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001244000.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001244000.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001244000.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001244000.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001244000.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001244000.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001244000.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001244000.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001244000.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_001244000.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_003121.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_003121.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_003121.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_003121.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_003121.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_003121.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_003121.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_003121.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_003121.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_003121.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_003121.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900011788.hg.1 NM_003121.4 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000270632.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000270632.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000270632.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000270632.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000270632.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000270632.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000270632.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000270632.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000270632.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000270632.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000270632.7 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000439922.6 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000439922.6 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000439922.6 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000439922.6 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000439922.6 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000439922.6 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000439922.6 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000439922.6 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000439922.6 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000439922.6 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000594188.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000594188.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000594188.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000594188.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000594188.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000594685.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000594685.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000594685.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000594685.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000594685.1 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000595883.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000595883.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000595883.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000595883.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000595883.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000595883.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000595883.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000595883.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000595883.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000595883.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000595883.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000595883.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000595883.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000596074.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000596074.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000596074.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000596074.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000596074.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000596074.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000596074.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000596074.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000597855.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000597855.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000597855.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000597855.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000597855.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000597855.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000597855.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000599923.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000599923.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000599923.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 ENST00000599923.5 Ensembl
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001243998.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001243998.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001243998.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001243998.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001243998.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001243998.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001243998.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001243998.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001243998.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001243998.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001243999.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001243999.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001243999.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001243999.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001243999.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001243999.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001243999.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001243999.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001243999.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001243999.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001243999.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001243999.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001244000.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001244000.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001244000.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001244000.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001244000.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001244000.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001244000.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001244000.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001244000.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001244000.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_001244000.1 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_003121.4 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_003121.4 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_003121.4 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_003121.4 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_003121.4 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_003121.4 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_003121.4 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_003121.4 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_003121.4 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_003121.4 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_003121.4 Refseq
Affymetrix Human Clariom D Assay TC1900011788.hg.1 NM_003121.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-859c6f577-lllsl:80/100.66.75.98:80.
git-commit: 13ffd1f44fd3b57634547fe5abf56598aa1cdbac
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.44.0-2026.02.07-1.0