This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01562948_m1
See other GPX8 GE Assays ›
Gene Symbol:
GPX8
Gene Name:
glutathione peroxidase 8 (putative)
Gene Aliases:
EPLA847, GPx-8, GSHPx-8, UNQ847
Chromosome Location:
Chr.5: 55160118 - 55167301 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01562948_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPX8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001008397.3 NP_001008398.2 2 - 3 536 93
NM_001306197.1 NP_001293126.1 2 - 3 383 93
NM_001306201.1 NP_001293130.1 2 - 3 257 93
NR_131336.1 3 - 4 450 93
NR_131337.1 3 - 4 392 93
NR_131340.1 2 - 3 526 93
XM_006714631.2 XP_006714694.1 2 - 3 502 93
GenBank mRNA AK074216.1 - 2 - 3 539 93
AK298545.1 - 2 - 3 331 93
AY358715.1 - 2 - 3 462 93
BC029424.1 - 2 - 3 495 4285958 93
BM835879.1 - 3 - 4 343 93
CN390753.1 - 2 - 3 323 93
DA660205.1 - 2 - 3 193 93

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPX8

Panther Classification:

Molecular Function -

peroxidase

Gene Ontology Categories:

Function(s) Process(es)

response to reactive oxygen species
oxidation-reduction process
cellular oxidant detoxification
peroxidase activity
glutathione peroxidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0