This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01565084_m1
See other PTPRD GE Assays ›
Gene Symbol:
PTPRD
Gene Name:
protein tyrosine phosphatase, receptor type D
Gene Aliases:
HPTP, HPTPD, HPTPDELTA, PTPD, RPTPDELTA
Chromosome Location:
Chr.9: 8314246 - 10612840 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01565084_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PTPRD
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006716827.3 XP_006716890.1 21 - 22 3314 86
XM_017014972.1 XP_016870461.1 24 - 25 3359 86
XM_017014974.1 XP_016870463.1 24 - 25 3359 86
XM_017014977.1 XP_016870466.1 24 - 25 3357 86
XM_017014980.1 XP_016870469.1 24 - 25 3353 86
XM_017014981.1 XP_016870470.1 21 - 22 3309 86

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
phosphate-containing compound metabolic process
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
transmembrane receptor protein tyrosine phosphatase signaling pathway
chemical synaptic transmission
neuron differentiation
peptidyl-tyrosine dephosphorylation
positive regulation of dendrite morphogenesis
presynaptic membrane assembly
transmembrane receptor protein tyrosine phosphatase activity
receptor binding
protein binding
cell adhesion molecule binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline