This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01565289_m1
See other NDUFS1 GE Assays ›
Gene Symbol:
NDUFS1
Gene Name:
NADH:ubiquinone oxidoreductase core subunit S1
Gene Aliases:
CI-75Kd, CI-75k, PRO1304
Chromosome Location:
Chr.2: 206121971 - 206159519 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01565289_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NDUFS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001199984.1 NP_001186913.1 2 - 3 170 147
GenBank mRNA AK295987.1 - 2 - 3 170 147

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial electron transport, NADH to ubiquinone
apoptotic mitochondrial changes
mitochondrial respiratory chain complex I assembly
cellular respiration
ATP metabolic process
regulation of mitochondrial membrane potential
reactive oxygen species metabolic process
protein binding
NADH dehydrogenase (ubiquinone) activity
electron carrier activity
metal ion binding
2 iron, 2 sulfur cluster binding
4 iron, 4 sulfur cluster binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0