This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01566651_m1
See other DUSP15 GE Assays ›
Gene Symbol:
DUSP15
Gene Name:
dual specificity phosphatase 15
Gene Aliases:
C20orf57, VHY
Chromosome Location:
Chr.20: 31845578 - 31870744 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01566651_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DUSP15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001012644.2 NP_001012662.1 3 - 4 547 153
NM_001320478.1 NP_001307407.1 3 - 4 204 153
NM_001320479.1 NP_001307408.1 3 - 4 204 153
NM_080611.4 NP_542178.2 3 - 4 543 153
NM_177991.2 NP_817130.1 3 - 4 309 153
XM_017027654.1 XP_016883143.1 3 - 4 401 153
XM_017027655.1 XP_016883144.1 3 - 4 192 153
XM_017027656.1 XP_016883145.1 3 - 4 401 153
XM_017027657.1 XP_016883146.1 3 - 4 401 153
XM_017027658.1 XP_016883147.1 3 - 4 341 153
XM_017027659.1 XP_016883148.1 3 - 4 347 153
XM_017027660.1 XP_016883149.1 3 - 4 229 153
XM_017027661.1 XP_016883150.1 2 - 3 338 153
XM_017027662.1 XP_016883151.1 3 - 4 199 153
GenBank mRNA AK091960.1 - 2 - 3 743 153
AK097430.1 - 3 - 4 206 153
BC056911.1 - 3 - 4 360 6195838 153
BF377396.1 - 3 - 4 153
BI517878.1 - 3 - 4 276 153
BI560652.1 - 3 - 4 547 153
BI562598.1 - 3 - 4 354 153
BI829823.1 - 3 - 4 474 153
BM554314.1 - 3 - 4 169 153
DN996690.1 - 3 - 4 316 153

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK091960.1, NM_001012644.2, NM_177991.2, XM_017027658.1, XM_017027659.1, XM_017027660.1, XM_017027661.1, XM_017027662.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DUSP15

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

transforming growth factor beta receptor signaling pathway
dephosphorylation
peptidyl-tyrosine dephosphorylation
regulation of cell proliferation
positive regulation of JNK cascade
protein tyrosine phosphatase activity
protein binding
protein tyrosine/serine/threonine phosphatase activity
phosphatase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000339738.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000339738.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000339738.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000339738.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000339738.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000339738.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000339738.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000339738.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000339738.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000339738.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000339738.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000339738.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000339738.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000375966.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000375966.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000375966.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000375966.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000375966.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000375966.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000375966.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000375966.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000375966.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000398083.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000398083.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000398083.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000398083.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000398083.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000398083.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000398083.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000398083.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000398083.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000398084.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000398084.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000398084.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000398084.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000398084.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000398084.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000398084.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000398084.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000398084.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000398084.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000428829.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000428829.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000428829.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000428829.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000428829.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000459848.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000459848.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000459848.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000459848.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000459848.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000459848.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000459848.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000459848.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000459848.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000486996.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000486996.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000486996.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000486996.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000486996.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000486996.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000486996.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000486996.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000486996.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000486996.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000486996.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000486996.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000493115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000493115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000493115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000493115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000493115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000493115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 ENST00000493115.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 NM_001012644.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 NM_001012644.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 NM_001012644.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 NM_001012644.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 NM_001012644.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 NM_001012644.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 NM_001012644.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 NM_001012644.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 NM_001012644.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 NM_080611.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 NM_080611.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 NM_080611.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 NM_080611.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 NM_080611.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 NM_080611.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 NM_080611.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 NM_080611.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 NM_080611.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 NM_080611.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 NM_080611.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 NM_080611.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 NM_177991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 NM_177991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 NM_177991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 NM_177991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 NM_177991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 NM_177991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 NM_177991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 NM_177991.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000009997.hg.1 NM_177991.2 Refseq
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000339738.9 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000339738.9 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000339738.9 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000339738.9 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000339738.9 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000339738.9 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000339738.9 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000339738.9 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000339738.9 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000339738.9 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000339738.9 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000339738.9 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000339738.9 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000375966.8 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000375966.8 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000375966.8 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000375966.8 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000375966.8 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000375966.8 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000375966.8 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000375966.8 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000375966.8 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000398083.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000398083.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000398083.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000398083.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000398083.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000398083.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000398083.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000398083.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000398083.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000398084.6 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000398084.6 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000398084.6 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000398084.6 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000398084.6 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000398084.6 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000398084.6 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000398084.6 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000398084.6 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000398084.6 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000428829.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000428829.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000428829.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000428829.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000428829.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000459848.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000459848.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000459848.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000459848.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000459848.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000459848.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000459848.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000459848.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000459848.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000486996.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000486996.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000486996.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000486996.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000486996.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000486996.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000486996.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000486996.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000486996.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000486996.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000486996.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000486996.5 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000493115.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000493115.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000493115.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000493115.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000493115.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000493115.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 ENST00000493115.1 Ensembl
Affymetrix Human Clariom D Assay TC2000009997.hg.1 NM_001012644.2 Refseq
Affymetrix Human Clariom D Assay TC2000009997.hg.1 NM_001012644.2 Refseq
Affymetrix Human Clariom D Assay TC2000009997.hg.1 NM_001012644.2 Refseq
Affymetrix Human Clariom D Assay TC2000009997.hg.1 NM_001012644.2 Refseq
Affymetrix Human Clariom D Assay TC2000009997.hg.1 NM_001012644.2 Refseq
Affymetrix Human Clariom D Assay TC2000009997.hg.1 NM_001012644.2 Refseq
Affymetrix Human Clariom D Assay TC2000009997.hg.1 NM_001012644.2 Refseq
Affymetrix Human Clariom D Assay TC2000009997.hg.1 NM_001012644.2 Refseq
Affymetrix Human Clariom D Assay TC2000009997.hg.1 NM_001012644.2 Refseq
Affymetrix Human Clariom D Assay TC2000009997.hg.1 NM_080611.4 Refseq
Affymetrix Human Clariom D Assay TC2000009997.hg.1 NM_080611.4 Refseq
Affymetrix Human Clariom D Assay TC2000009997.hg.1 NM_080611.4 Refseq
Affymetrix Human Clariom D Assay TC2000009997.hg.1 NM_080611.4 Refseq
Affymetrix Human Clariom D Assay TC2000009997.hg.1 NM_080611.4 Refseq
Affymetrix Human Clariom D Assay TC2000009997.hg.1 NM_080611.4 Refseq
Affymetrix Human Clariom D Assay TC2000009997.hg.1 NM_080611.4 Refseq
Affymetrix Human Clariom D Assay TC2000009997.hg.1 NM_080611.4 Refseq
Affymetrix Human Clariom D Assay TC2000009997.hg.1 NM_080611.4 Refseq
Affymetrix Human Clariom D Assay TC2000009997.hg.1 NM_080611.4 Refseq
Affymetrix Human Clariom D Assay TC2000009997.hg.1 NM_080611.4 Refseq
Affymetrix Human Clariom D Assay TC2000009997.hg.1 NM_080611.4 Refseq
Affymetrix Human Clariom D Assay TC2000009997.hg.1 NM_177991.2 Refseq
Affymetrix Human Clariom D Assay TC2000009997.hg.1 NM_177991.2 Refseq
Affymetrix Human Clariom D Assay TC2000009997.hg.1 NM_177991.2 Refseq
Affymetrix Human Clariom D Assay TC2000009997.hg.1 NM_177991.2 Refseq
Affymetrix Human Clariom D Assay TC2000009997.hg.1 NM_177991.2 Refseq
Affymetrix Human Clariom D Assay TC2000009997.hg.1 NM_177991.2 Refseq
Affymetrix Human Clariom D Assay TC2000009997.hg.1 NM_177991.2 Refseq
Affymetrix Human Clariom D Assay TC2000009997.hg.1 NM_177991.2 Refseq
Affymetrix Human Clariom D Assay TC2000009997.hg.1 NM_177991.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline