This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01566856_m1
See other HDHD3 GE Assays ›
Gene Symbol:
HDHD3
Gene Name:
haloacid dehalogenase like hydrolase domain containing 3
Gene Aliases:
2810435D12Rik, C9orf158
Chromosome Location:
Chr.9: 113373416 - 113377009 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01566856_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HDHD3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001304509.1 NP_001291438.1 1 - 2 278 186
GenBank mRNA AK315401.1 - 1 - 2 222 186

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900011259.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK315401.1,NM_001304509.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
protein binding
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 ENST00000238379.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 ENST00000238379.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 ENST00000238379.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 ENST00000238379.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 ENST00000238379.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 ENST00000238379.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 ENST00000238379.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 ENST00000238379.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 ENST00000374180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 ENST00000374180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 ENST00000374180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 ENST00000374180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 ENST00000374180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 ENST00000374180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 ENST00000374180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 ENST00000374180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 ENST00000374180.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 ENST00000485934.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 ENST00000485934.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 ENST00000485934.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 ENST00000485934.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_001304509.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_001304509.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_001304509.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_001304509.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_001304509.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_001304509.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_001304509.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_001304509.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_001304509.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_001304510.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_001304510.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_001304510.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_001304510.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_001304510.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_001304510.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_001304510.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_001304510.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_001304511.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_001304511.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_001304511.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_001304511.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_001304511.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_001304511.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_001304511.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_031219.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_031219.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_031219.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_031219.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_031219.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_031219.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011259.hg.1 NM_031219.3 Refseq
Affymetrix Human Clariom D Assay PSR0900176742.hg.1 ENST00000238379.9 Ensembl
Affymetrix Human Clariom D Assay PSR0900176742.hg.1 ENST00000374180.3 Ensembl
Affymetrix Human Clariom D Assay PSR0900176742.hg.1 NM_001304509.1 Refseq
Affymetrix Human Clariom D Assay PSR0900176742.hg.1 NM_031219.3 Refseq
Affymetrix Human Clariom D Assay PSR0900176742.hg.1 ENST00000238379.9 Ensembl
Affymetrix Human Clariom D Assay PSR0900176742.hg.1 ENST00000374180.3 Ensembl
Affymetrix Human Clariom D Assay PSR0900176742.hg.1 NM_001304509.1 Refseq
Affymetrix Human Clariom D Assay PSR0900176742.hg.1 NM_031219.3 Refseq
Affymetrix Human Clariom D Assay PSR0900176742.hg.1 TC0900011259.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0900176746.hg.1 ENST00000374180.3 Ensembl
Affymetrix Human Clariom D Assay PSR0900176746.hg.1 ENST00000485934.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900176746.hg.1 NM_001304509.1 Refseq
Affymetrix Human Clariom D Assay PSR0900176746.hg.1 NM_001304510.1 Refseq
Affymetrix Human Clariom D Assay PSR0900176746.hg.1 NM_001304511.1 Refseq
Affymetrix Human Clariom D Assay PSR0900176746.hg.1 ENST00000374180.3 Ensembl
Affymetrix Human Clariom D Assay PSR0900176746.hg.1 ENST00000485934.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900176746.hg.1 NM_001304509.1 Refseq
Affymetrix Human Clariom D Assay PSR0900176746.hg.1 NM_001304510.1 Refseq
Affymetrix Human Clariom D Assay PSR0900176746.hg.1 NM_001304511.1 Refseq
Affymetrix Human Clariom D Assay PSR0900176746.hg.1 TC0900011259.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0900176747.hg.1 ENST00000374180.3 Ensembl
Affymetrix Human Clariom D Assay PSR0900176747.hg.1 NM_001304509.1 Refseq
Affymetrix Human Clariom D Assay PSR0900176747.hg.1 NM_001304510.1 Refseq
Affymetrix Human Clariom D Assay PSR0900176747.hg.1 NM_001304511.1 Refseq
Affymetrix Human Clariom D Assay PSR0900176747.hg.1 ENST00000374180.3 Ensembl
Affymetrix Human Clariom D Assay PSR0900176747.hg.1 NM_001304509.1 Refseq
Affymetrix Human Clariom D Assay PSR0900176747.hg.1 NM_001304510.1 Refseq
Affymetrix Human Clariom D Assay PSR0900176747.hg.1 NM_001304511.1 Refseq
Affymetrix Human Clariom D Assay PSR0900176747.hg.1 TC0900011259.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0900011259.hg.1 ENST00000238379.9 Ensembl
Affymetrix Human Clariom D Assay TC0900011259.hg.1 ENST00000238379.9 Ensembl
Affymetrix Human Clariom D Assay TC0900011259.hg.1 ENST00000238379.9 Ensembl
Affymetrix Human Clariom D Assay TC0900011259.hg.1 ENST00000238379.9 Ensembl
Affymetrix Human Clariom D Assay TC0900011259.hg.1 ENST00000238379.9 Ensembl
Affymetrix Human Clariom D Assay TC0900011259.hg.1 ENST00000238379.9 Ensembl
Affymetrix Human Clariom D Assay TC0900011259.hg.1 ENST00000238379.9 Ensembl
Affymetrix Human Clariom D Assay TC0900011259.hg.1 ENST00000238379.9 Ensembl
Affymetrix Human Clariom D Assay TC0900011259.hg.1 ENST00000374180.3 Ensembl
Affymetrix Human Clariom D Assay TC0900011259.hg.1 ENST00000374180.3 Ensembl
Affymetrix Human Clariom D Assay TC0900011259.hg.1 ENST00000374180.3 Ensembl
Affymetrix Human Clariom D Assay TC0900011259.hg.1 ENST00000374180.3 Ensembl
Affymetrix Human Clariom D Assay TC0900011259.hg.1 ENST00000374180.3 Ensembl
Affymetrix Human Clariom D Assay TC0900011259.hg.1 ENST00000374180.3 Ensembl
Affymetrix Human Clariom D Assay TC0900011259.hg.1 ENST00000374180.3 Ensembl
Affymetrix Human Clariom D Assay TC0900011259.hg.1 ENST00000374180.3 Ensembl
Affymetrix Human Clariom D Assay TC0900011259.hg.1 ENST00000374180.3 Ensembl
Affymetrix Human Clariom D Assay TC0900011259.hg.1 ENST00000485934.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011259.hg.1 ENST00000485934.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011259.hg.1 ENST00000485934.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011259.hg.1 ENST00000485934.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_001304509.1 Refseq
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_001304509.1 Refseq
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_001304509.1 Refseq
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_001304509.1 Refseq
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_001304509.1 Refseq
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_001304509.1 Refseq
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_001304509.1 Refseq
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_001304509.1 Refseq
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_001304509.1 Refseq
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_001304510.1 Refseq
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_001304510.1 Refseq
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_001304510.1 Refseq
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_001304510.1 Refseq
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_001304510.1 Refseq
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_001304510.1 Refseq
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_001304510.1 Refseq
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_001304510.1 Refseq
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_001304511.1 Refseq
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_001304511.1 Refseq
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_001304511.1 Refseq
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_001304511.1 Refseq
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_001304511.1 Refseq
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_001304511.1 Refseq
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_001304511.1 Refseq
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_031219.3 Refseq
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_031219.3 Refseq
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_031219.3 Refseq
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_031219.3 Refseq
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_031219.3 Refseq
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_031219.3 Refseq
Affymetrix Human Clariom D Assay TC0900011259.hg.1 NM_031219.3 Refseq
Affymetrix Human Clariom D Assay PSR0900176748.hg.1 ENST00000374180.3 Ensembl
Affymetrix Human Clariom D Assay PSR0900176748.hg.1 NM_001304509.1 Refseq
Affymetrix Human Clariom D Assay PSR0900176748.hg.1 NM_001304510.1 Refseq
Affymetrix Human Clariom D Assay PSR0900176748.hg.1 NM_001304511.1 Refseq
Affymetrix Human Clariom D Assay PSR0900176748.hg.1 ENST00000374180.3 Ensembl
Affymetrix Human Clariom D Assay PSR0900176748.hg.1 NM_001304509.1 Refseq
Affymetrix Human Clariom D Assay PSR0900176748.hg.1 NM_001304510.1 Refseq
Affymetrix Human Clariom D Assay PSR0900176748.hg.1 NM_001304511.1 Refseq
Affymetrix Human Clariom D Assay PSR0900176748.hg.1 TC0900011259.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline