| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000355904.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000355904.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000355904.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000355904.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000355904.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000355904.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000355904.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000355904.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000355904.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000355904.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000355904.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000355904.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000355904.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000355904.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000518445.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000518445.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000518445.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000518445.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000518599.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000518599.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000518599.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000518599.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000518599.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000518599.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000518599.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000522833.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000522833.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000522833.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000522833.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000522833.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000523499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000523499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000523499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000523499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000523499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000523499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000523499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000523499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000523499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
ENST00000523499.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
NM_002095.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
NM_002095.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
NM_002095.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
NM_002095.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
NM_002095.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
NM_002095.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
NM_002095.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
NM_002095.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
NM_002095.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
NM_002095.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
NM_002095.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800012404.hg.1 |
NM_002095.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08014296.hg.1 |
ENST00000355904.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08014296.hg.1 |
NM_002095.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08014296.hg.1 |
TC08001109.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0800186939.hg.1 |
ENST00000355904.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800186939.hg.1 |
ENST00000518445.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800186939.hg.1 |
ENST00000518599.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800186939.hg.1 |
ENST00000523499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800186939.hg.1 |
NM_002095.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800186939.hg.1 |
ENST00000355904.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800186939.hg.1 |
ENST00000518445.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800186939.hg.1 |
ENST00000518599.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800186939.hg.1 |
ENST00000523499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800186939.hg.1 |
NM_002095.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800186939.hg.1 |
TC0800012404.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000355904.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000355904.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000355904.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000355904.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000355904.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000355904.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000355904.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000355904.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000355904.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000355904.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000355904.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000355904.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000355904.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000355904.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000518445.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000518445.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000518445.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000518445.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000518599.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000518599.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000518599.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000518599.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000518599.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000518599.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000518599.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000522833.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000522833.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000522833.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000522833.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000522833.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000523499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000523499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000523499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000523499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000523499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000523499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000523499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000523499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000523499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
ENST00000523499.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
NM_002095.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
NM_002095.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
NM_002095.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
NM_002095.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
NM_002095.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
NM_002095.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
NM_002095.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
NM_002095.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
NM_002095.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
NM_002095.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
NM_002095.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800012404.hg.1 |
NM_002095.5 |
Refseq |