This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01568682_m1
See other GTF2E2 GE Assays ›
Gene Symbol:
GTF2E2
Gene Name:
general transcription factor IIE subunit 2
Gene Aliases:
FE, TF2E2, TFIIE-B, TTD6
Chromosome Location:
Chr.8: 30578514 - 30658241 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01568682_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GTF2E2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002095.4 NP_002086.1 3 - 4 514 152
XM_011544509.2 XP_011542811.1 3 - 4 534 152
XM_011544510.1 XP_011542812.1 3 - 4 534 152
XM_017013363.1 XP_016868852.1 3 - 4 374 152
XM_017013364.1 XP_016868853.1 3 - 4 304 152
GenBank mRNA BC030572.2 - 3 - 4 508 3955818 152
S67861.1 - 3 - 4 451 152
X63469.1 - 3 - 4 503 152

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0800012404.hg.1
This assay targets the same location as HTA transcript cluster(s) TC08001109.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GTF2E2

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
snRNA transcription from RNA polymerase II promoter
DNA binding
protein binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR08014295.hg.1 ENST00000355904.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR08014295.hg.1 NM_002095.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR08014295.hg.1 TC08001109.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000355904.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000355904.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000355904.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000355904.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000355904.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000355904.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000355904.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000355904.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000355904.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000355904.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000355904.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000355904.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000355904.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000355904.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000518445.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000518445.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000518445.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000518445.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000518599.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000518599.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000518599.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000518599.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000518599.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000518599.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000518599.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000522833.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000522833.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000522833.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000522833.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000522833.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000523499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000523499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000523499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000523499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000523499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000523499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000523499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000523499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000523499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 ENST00000523499.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 NM_002095.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 NM_002095.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 NM_002095.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 NM_002095.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 NM_002095.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 NM_002095.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 NM_002095.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 NM_002095.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 NM_002095.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 NM_002095.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 NM_002095.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012404.hg.1 NM_002095.5 Refseq
Affymetrix Human Clariom D Assay PSR0800186938.hg.1 ENST00000355904.8 Ensembl
Affymetrix Human Clariom D Assay PSR0800186938.hg.1 ENST00000518445.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800186938.hg.1 ENST00000518599.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800186938.hg.1 ENST00000523499.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800186938.hg.1 NM_002095.5 Refseq
Affymetrix Human Clariom D Assay PSR0800186938.hg.1 ENST00000355904.8 Ensembl
Affymetrix Human Clariom D Assay PSR0800186938.hg.1 ENST00000518445.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800186938.hg.1 ENST00000518599.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800186938.hg.1 ENST00000523499.5 Ensembl
Affymetrix Human Clariom D Assay PSR0800186938.hg.1 NM_002095.5 Refseq
Affymetrix Human Clariom D Assay PSR0800186938.hg.1 TC0800012404.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000355904.8 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000355904.8 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000355904.8 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000355904.8 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000355904.8 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000355904.8 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000355904.8 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000355904.8 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000355904.8 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000355904.8 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000355904.8 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000355904.8 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000355904.8 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000355904.8 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000518445.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000518445.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000518445.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000518445.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000518599.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000518599.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000518599.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000518599.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000518599.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000518599.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000518599.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000522833.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000522833.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000522833.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000522833.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000522833.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000523499.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000523499.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000523499.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000523499.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000523499.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000523499.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000523499.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000523499.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000523499.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 ENST00000523499.5 Ensembl
Affymetrix Human Clariom D Assay TC0800012404.hg.1 NM_002095.5 Refseq
Affymetrix Human Clariom D Assay TC0800012404.hg.1 NM_002095.5 Refseq
Affymetrix Human Clariom D Assay TC0800012404.hg.1 NM_002095.5 Refseq
Affymetrix Human Clariom D Assay TC0800012404.hg.1 NM_002095.5 Refseq
Affymetrix Human Clariom D Assay TC0800012404.hg.1 NM_002095.5 Refseq
Affymetrix Human Clariom D Assay TC0800012404.hg.1 NM_002095.5 Refseq
Affymetrix Human Clariom D Assay TC0800012404.hg.1 NM_002095.5 Refseq
Affymetrix Human Clariom D Assay TC0800012404.hg.1 NM_002095.5 Refseq
Affymetrix Human Clariom D Assay TC0800012404.hg.1 NM_002095.5 Refseq
Affymetrix Human Clariom D Assay TC0800012404.hg.1 NM_002095.5 Refseq
Affymetrix Human Clariom D Assay TC0800012404.hg.1 NM_002095.5 Refseq
Affymetrix Human Clariom D Assay TC0800012404.hg.1 NM_002095.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline