Assay Details
| Gene Symbol: GTF2E2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002095.4 | NP_002086.1 | 6 - 7 | 895 | 148 | |
| XM_017013363.1 | XP_016868852.1 | 6 - 7 | 755 | 148 | ||
| XM_017013364.1 | XP_016868853.1 | 6 - 7 | 685 | 148 | ||
| GenBank mRNA | BC030572.2 | - | 6 - 7 | 889 | 3955818 | 148 |
| S67861.1 | - | 6 - 7 | 832 | 148 | ||
| X63469.1 | - | 6 - 7 | 884 | 148 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001109.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
snRNA transcription from RNA polymerase II promoter
DNA binding
protein binding
poly(A) RNA binding
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
snRNA transcription from RNA polymerase II promoter
DNA binding
protein binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08014291.hg.1 | ENST00000355904.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014291.hg.1 | ENST00000522833.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014291.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014291.hg.1 | TC08001109.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014287.hg.1 | ENST00000355904.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014287.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014287.hg.1 | TC08001109.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000518445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000518445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000518445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000518445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000518599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000518599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000518599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000518599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000518599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000518599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000518599.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000522833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000522833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000522833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000522833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000522833.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000523499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000523499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000523499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000523499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000523499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000523499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000523499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000523499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000523499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | ENST00000523499.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012404.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014290.hg.1 | ENST00000355904.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014290.hg.1 | ENST00000522833.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014290.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08014290.hg.1 | TC08001109.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0800186930.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186930.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800186930.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186930.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800186930.hg.1 | TC0800012404.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800186934.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186934.hg.1 | ENST00000518599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186934.hg.1 | ENST00000522833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186934.hg.1 | ENST00000523499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186934.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800186934.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186934.hg.1 | ENST00000518599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186934.hg.1 | ENST00000522833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186934.hg.1 | ENST00000523499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186934.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800186934.hg.1 | TC0800012404.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000518445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000518445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000518445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000518445.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000518599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000518599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000518599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000518599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000518599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000518599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000518599.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000522833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000522833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000522833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000522833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000522833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000523499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000523499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000523499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000523499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000523499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000523499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000523499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000523499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000523499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | ENST00000523499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012404.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800186933.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186933.hg.1 | ENST00000522833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186933.hg.1 | ENST00000523499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186933.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800186933.hg.1 | ENST00000355904.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186933.hg.1 | ENST00000522833.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186933.hg.1 | ENST00000523499.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186933.hg.1 | NM_002095.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800186933.hg.1 | TC0800012404.hg.1 | Clariom D TC |

GTF2E2
Genomic Map