This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01569096_g1
See other RBM26 GE Assays ›
Gene Symbol:
RBM26
Gene Name:
RNA binding motif protein 26
Gene Aliases:
ARRS2, C13orf10, PPP1R132, PRO1777, SE70-2, ZC3H17
Chromosome Location:
Chr.13: 79311827 - 79406265 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01569096_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBM26
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK027772.1 - 1 846 203

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0