This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01570492_m1
See other FAT4 GE Assays ›
Gene Symbol:
FAT4
Gene Name:
FAT atypical cadherin 4
Gene Aliases:
CDHF14, CDHR11, FAT-J, FATJ, HKLLS2, NBLA00548, VMLDS2
Chromosome Location:
Chr.4: 125314913 - 125492932 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01570492_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FAT4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001291285.1 NP_001278214.1 13 - 14 12474 72
NM_001291303.1 NP_001278232.1 13 - 14 12474 72
NM_024582.4 NP_078858.4 13 - 14 12468 72
XM_011532236.2 XP_011530538.1 14 - 15 13551 72
XM_011532237.2 XP_011530539.1 13 - 14 8359 72
GenBank mRNA AK096646.1 - 5 - 6 770 72
AK291461.1 - 5 - 6 1014 72
KJ131486.1 - 5 - 6 711 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400008641.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04000636.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene FAT4

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

branching involved in ureteric bud morphogenesis
heart morphogenesis
plasma membrane organization
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
cerebral cortex development
neurogenesis
hippo signaling
ossification involved in bone maturation
digestive tract development
inner ear receptor stereocilium organization
regulation of metanephric nephron tubule epithelial cell differentiation
calcium ion binding
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR04009213.hg.1 ENST00000394329.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04009213.hg.1 NM_024582.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04009213.hg.1 TC04000636.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000509444.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000509444.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 ENST00000509444.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Clariom D Assay PSR0400159480.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Clariom D Assay PSR0400159480.hg.1 NM_001291285.1 Refseq
Affymetrix Human Clariom D Assay PSR0400159480.hg.1 NM_001291303.1 Refseq
Affymetrix Human Clariom D Assay PSR0400159480.hg.1 NM_024582.4 Refseq
Affymetrix Human Clariom D Assay PSR0400159480.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Clariom D Assay PSR0400159480.hg.1 NM_001291285.1 Refseq
Affymetrix Human Clariom D Assay PSR0400159480.hg.1 NM_001291303.1 Refseq
Affymetrix Human Clariom D Assay PSR0400159480.hg.1 NM_024582.4 Refseq
Affymetrix Human Clariom D Assay PSR0400159480.hg.1 TC0400008641.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000335110.5 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000394329.7 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000509444.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000509444.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 ENST00000509444.1 Ensembl
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291285.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_001291303.1 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_024582.4 Refseq
Affymetrix Human Clariom D Assay TC0400008641.hg.1 NM_024582.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline