Assay Details
| Gene Symbol: FAT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291285.1 | NP_001278214.1 | 7 - 8 | 7204 | 71 | |
| NM_001291303.1 | NP_001278232.1 | 7 - 8 | 7204 | 71 | ||
| XM_011532236.2 | XP_011530538.1 | 8 - 9 | 8281 | 71 | ||
| XM_011532237.2 | XP_011530539.1 | 7 - 8 | 3089 | 71 | ||
| GenBank mRNA | KJ131481.1 | - | 1 - 2 | 81 | 71 | |
| KJ131482.1 | - | 1 - 2 | 88 | 71 | ||
| KJ131485.1 | - | 3 - 4 | 388 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
branching involved in ureteric bud morphogenesis
heart morphogenesis
plasma membrane organization
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
cerebral cortex development
neurogenesis
hippo signaling
ossification involved in bone maturation
digestive tract development
inner ear receptor stereocilium organization
regulation of metanephric nephron tubule epithelial cell differentiation
calcium ion binding
protein binding
branching involved in ureteric bud morphogenesis
heart morphogenesis
plasma membrane organization
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
cerebral cortex development
neurogenesis
hippo signaling
ossification involved in bone maturation
digestive tract development
inner ear receptor stereocilium organization
regulation of metanephric nephron tubule epithelial cell differentiation
calcium ion binding
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000509444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000509444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | ENST00000509444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400159468.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400159468.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400159468.hg.1 | ENST00000509444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400159468.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400159468.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400159468.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400159468.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400159468.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400159468.hg.1 | ENST00000509444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400159468.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400159468.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400159468.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400159468.hg.1 | TC0400008641.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000335110.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000394329.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000509444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000509444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | ENST00000509444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291285.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_001291303.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_024582.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400008641.hg.1 | NM_024582.4 | Refseq |

FAT4
Genomic Map