This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01573162_m1
See other FABP2 GE Assays ›
Gene Symbol:
FABP2
Gene Name:
fatty acid binding protein 2
Gene Aliases:
FABPI, I-FABP
Chromosome Location:
Chr.4: 119317250 - 119322161 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01573162_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FABP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000134.3 NP_000125.2 1 - 2 122 88
GenBank mRNA AK025079.1 - 1 - 2 95 88
BC069466.1 - 1 - 2 113 7262448 88
BC069617.1 - 1 - 2 106 7262424 88
BC069625.1 - 1 - 2 113 7262436 88
BC069637.1 - 1 - 2 113 7262460 88
BC111791.1 - 1 - 2 122 40017872 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400011706.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04001500.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene FABP2

Gene Ontology Categories:

Function(s) Process(es)

transport
digestion
triglyceride catabolic process
transporter activity
fatty acid binding
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR04021276.hg.1 TC04001500.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04021276.hg.1 ENST00000274024.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04021276.hg.1 NM_000134.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011706.hg.1 ENST00000274024.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011706.hg.1 ENST00000274024.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011706.hg.1 ENST00000274024.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011706.hg.1 ENST00000274024.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011706.hg.1 ENST00000274024.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011706.hg.1 ENST00000274024.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011706.hg.1 ENST00000274024.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011706.hg.1 ENST00000274024.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011706.hg.1 ENST00000274024.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400011706.hg.1 NM_000134.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011706.hg.1 NM_000134.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011706.hg.1 NM_000134.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011706.hg.1 NM_000134.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011706.hg.1 NM_000134.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011706.hg.1 NM_000134.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011706.hg.1 NM_000134.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400011706.hg.1 NM_000134.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04021275.hg.1 TC04001500.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04021275.hg.1 ENST00000274024.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04021275.hg.1 NM_000134.3 Refseq
Affymetrix Human Clariom D Assay PSR0400180423.hg.1 ENST00000274024.3 Ensembl
Affymetrix Human Clariom D Assay PSR0400180423.hg.1 NM_000134.3 Refseq
Affymetrix Human Clariom D Assay PSR0400180423.hg.1 ENST00000274024.3 Ensembl
Affymetrix Human Clariom D Assay PSR0400180423.hg.1 NM_000134.3 Refseq
Affymetrix Human Clariom D Assay PSR0400180423.hg.1 TC0400011706.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0400011706.hg.1 ENST00000274024.3 Ensembl
Affymetrix Human Clariom D Assay TC0400011706.hg.1 ENST00000274024.3 Ensembl
Affymetrix Human Clariom D Assay TC0400011706.hg.1 ENST00000274024.3 Ensembl
Affymetrix Human Clariom D Assay TC0400011706.hg.1 ENST00000274024.3 Ensembl
Affymetrix Human Clariom D Assay TC0400011706.hg.1 ENST00000274024.3 Ensembl
Affymetrix Human Clariom D Assay TC0400011706.hg.1 ENST00000274024.3 Ensembl
Affymetrix Human Clariom D Assay TC0400011706.hg.1 ENST00000274024.3 Ensembl
Affymetrix Human Clariom D Assay TC0400011706.hg.1 ENST00000274024.3 Ensembl
Affymetrix Human Clariom D Assay TC0400011706.hg.1 ENST00000274024.3 Ensembl
Affymetrix Human Clariom D Assay TC0400011706.hg.1 NM_000134.3 Refseq
Affymetrix Human Clariom D Assay TC0400011706.hg.1 NM_000134.3 Refseq
Affymetrix Human Clariom D Assay TC0400011706.hg.1 NM_000134.3 Refseq
Affymetrix Human Clariom D Assay TC0400011706.hg.1 NM_000134.3 Refseq
Affymetrix Human Clariom D Assay TC0400011706.hg.1 NM_000134.3 Refseq
Affymetrix Human Clariom D Assay TC0400011706.hg.1 NM_000134.3 Refseq
Affymetrix Human Clariom D Assay TC0400011706.hg.1 NM_000134.3 Refseq
Affymetrix Human Clariom D Assay TC0400011706.hg.1 NM_000134.3 Refseq
Affymetrix Human Clariom D Assay PSR0400180422.hg.1 ENST00000274024.3 Ensembl
Affymetrix Human Clariom D Assay PSR0400180422.hg.1 NM_000134.3 Refseq
Affymetrix Human Clariom D Assay PSR0400180422.hg.1 ENST00000274024.3 Ensembl
Affymetrix Human Clariom D Assay PSR0400180422.hg.1 NM_000134.3 Refseq
Affymetrix Human Clariom D Assay PSR0400180422.hg.1 TC0400011706.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0400180424.hg.1 ENST00000274024.3 Ensembl
Affymetrix Human Clariom D Assay PSR0400180424.hg.1 NM_000134.3 Refseq
Affymetrix Human Clariom D Assay PSR0400180424.hg.1 ENST00000274024.3 Ensembl
Affymetrix Human Clariom D Assay PSR0400180424.hg.1 NM_000134.3 Refseq
Affymetrix Human Clariom D Assay PSR0400180424.hg.1 TC0400011706.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0