This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01573889_g1
See other SOAT2 GE Assays ›
Gene Symbol:
SOAT2
Gene Name:
sterol O-acyltransferase 2
Gene Aliases:
ACACT2, ACAT2, ARGP2
Chromosome Location:
Chr.12: 53102437 - 53124539 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01573889_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SOAT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003578.3 NP_003569.1 9 - 10 999 61
XM_011538849.2 XP_011537151.1 8 - 9 1992 61
XM_011538850.2 XP_011537152.1 7 - 8 1884 61
GenBank mRNA AF059203.1 - 9 - 10 962 61
AF099031.2 - 9 - 10 911 61
BC096090.1 - 9 - 10 959 40000865 61
BC096091.3 - 9 - 10 959 40000866 61
BC096092.3 - 9 - 10 959 40000868 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200007694.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12000429.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SOAT2

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

cholesterol metabolic process
macrophage derived foam cell differentiation
intestinal cholesterol absorption
cholesterol efflux
very-low-density lipoprotein particle assembly
cholesterol esterification
cholesterol homeostasis
fatty-acyl-CoA binding
cholesterol binding
transferase activity, transferring acyl groups
cholesterol O-acyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000551896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000551896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000551896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000551896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000551896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 ENST00000551896.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12005545.hg.1 ENST00000301466.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12005545.hg.1 NM_003578.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12005545.hg.1 TC12000429.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1200155000.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200155000.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200155000.hg.1 NM_003578.3 Refseq
Affymetrix Human Clariom D Assay PSR1200155000.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200155000.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200155000.hg.1 NM_003578.3 Refseq
Affymetrix Human Clariom D Assay PSR1200155000.hg.1 TC1200007694.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000301466.7 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000542365.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000551896.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000551896.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000551896.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000551896.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000551896.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 ENST00000551896.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Clariom D Assay TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Clariom D Assay TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Clariom D Assay TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Clariom D Assay TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Clariom D Assay TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Clariom D Assay TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Clariom D Assay TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Clariom D Assay TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Clariom D Assay TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Clariom D Assay TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Clariom D Assay TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Clariom D Assay TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Clariom D Assay TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Clariom D Assay TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Clariom D Assay TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Clariom D Assay TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Clariom D Assay TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Clariom D Assay TC1200007694.hg.1 NM_003578.3 Refseq
Affymetrix Human Clariom D Assay TC1200007694.hg.1 NM_003578.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0