This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01574496_g1
See other CLCN2 GE Assays ›
Gene Symbol:
CLCN2
Gene Name:
chloride voltage-gated channel 2
Gene Aliases:
CIC-2, CLC2, ECA2, ECA3, EGI11, EGI3, EGMA, EJM6, EJM8, LKPAT, clC-2
Chromosome Location:
Chr.3: 184346185 - 184361651 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01574496_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CLCN2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001171087.2 NP_001164558.1 21 - 22 2428 97
NM_001171088.2 NP_001164559.1 20 - 21 2347 97
NM_001171089.2 NP_001164560.1 21 - 22 2479 97
NM_004366.5 NP_004357.3 21 - 22 2479 97
XM_006713490.2 XP_006713553.1 15 - 16 1689 97
XM_011512401.1 XP_011510703.1 21 - 22 2431 97
GenBank mRNA AF026004.1 - 21 - 22 2307 97
AK298952.1 - 21 - 22 2431 97
AK302759.1 - 20 - 21 2299 97
BC072004.1 - 21 - 22 2379 6424015 97
BX089717.1 - 4 - 5 289 97
KU177982.1 - 8 - 9 879 97
KU177983.1 - 8 - 9 873 97
KU177984.1 - 8 - 9 807 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300013384.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CLCN2

Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transmembrane transport
retina development in camera-type eye
cell differentiation involved in salivary gland development
chloride transmembrane transport
regulation of anion transmembrane transport
voltage-gated chloride channel activity
chloride ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR0300199783.hg.1 ENST00000265593.8 Ensembl
Affymetrix Human Clariom D Assay PSR0300199783.hg.1 ENST00000344937.11 Ensembl
Affymetrix Human Clariom D Assay PSR0300199783.hg.1 ENST00000434054.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300199783.hg.1 ENST00000457512.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300199783.hg.1 NM_001171087.2 Refseq
Affymetrix Human Clariom D Assay PSR0300199783.hg.1 NM_001171088.2 Refseq
Affymetrix Human Clariom D Assay PSR0300199783.hg.1 NM_001171089.2 Refseq
Affymetrix Human Clariom D Assay PSR0300199783.hg.1 NM_004366.5 Refseq
Affymetrix Human Clariom D Assay PSR0300199783.hg.1 ENST00000265593.8 Ensembl
Affymetrix Human Clariom D Assay PSR0300199783.hg.1 ENST00000344937.11 Ensembl
Affymetrix Human Clariom D Assay PSR0300199783.hg.1 ENST00000434054.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300199783.hg.1 ENST00000457512.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300199783.hg.1 NM_001171087.2 Refseq
Affymetrix Human Clariom D Assay PSR0300199783.hg.1 NM_001171088.2 Refseq
Affymetrix Human Clariom D Assay PSR0300199783.hg.1 NM_001171089.2 Refseq
Affymetrix Human Clariom D Assay PSR0300199783.hg.1 NM_004366.5 Refseq
Affymetrix Human Clariom D Assay PSR0300199783.hg.1 TC0300013384.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline