This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01586663_s1
See other LOC728868 GE Assays ›
Gene Symbol:
LOC728868
Gene Name:
uncharacterized LOC728868
Gene Aliases:
-
Chromosome Location:
-
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01586663_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC728868
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK096660.1 - 1 2216 71
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB326331.1 - 1 Chr. 8 - 10423908 - 10424444 [+] on Build GRCh38 71

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 124-1 MIR124-1 MIR124A,
MIR124A1,
MIRN124-1,
MIRN124A1,
mir-124-1
-520.4kb Anti-sense Chr. 8 - 9903388 - 9903472 on NCBI Build GRCh38
Downstream uncharacterized LINCR-0001 LINCR-0001 50.1kb Sense Chr. 8 - 10474565 - 10481974 on NCBI Build GRCh38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3614348 - -
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 15803 - -
SurePrint G3 Human GE Microarray G4851A A_33_P3614348 - -
Array Name Probe IDs Targets Target Type
Affymetrix HT HG-U133+ PM Array Plate 1559586_PM_at - -
Human Genome U133 Plus 2.0 Array 1559586_at - -
Human Genome U133 Plus 2.0 Array 1559586_PM_at - -
Affymetrix Human Genome U133 Plus 2.0 Array 1559586_at8 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1559586_at9 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1559586_at4 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1559586_at1 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1559586_at2 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1559586_at11 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1559586_at10 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1559586_at7 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1559586_at3 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1559586_at5 - -
Affymetrix Human Genome U133 Plus 2.0 Array 1559586_at6 - -
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 235403 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline