This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01588916_g1
See other RPS9 GE Assays ›
Gene Symbol:
RPS9
Gene Name:
ribosomal protein S9
Gene Aliases:
S9
Chromosome Location:
Chr.19: 54200557 - 54207647 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01588916_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPS9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017027112.1 XP_016882601.1 1 - 2 340 148

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017027112.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
positive regulation of cell proliferation
viral transcription
positive regulation of translational fidelity
structural constituent of ribosome
protein binding
rRNA binding
poly(A) RNA binding
translation regulator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000302907.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000302907.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000302907.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000302907.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000302907.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000302907.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000302907.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000302907.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000302907.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000302907.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000391751.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000391751.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000391751.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000391751.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000391751.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000391751.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000391751.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000391752.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000391752.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000391752.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000391752.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000391752.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000391752.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000391752.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000391752.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000391752.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000391753.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000391753.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000391753.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000391753.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000391753.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000391753.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000391753.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000391753.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000402367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000402367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000402367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000402367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000402367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000402367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000402367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000402367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000402367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000402367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000402367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000402367.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000436445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000436445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000436445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000436445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000436445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000436445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000436445.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000441429.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000441429.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000441429.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000441429.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000441429.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000441429.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000441429.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000441429.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000441429.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000441429.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000441429.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000441429.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000445961.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000445961.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000445961.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000445961.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000445961.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000445961.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000445961.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000445961.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000445961.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000445961.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000448962.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000448962.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000448962.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000448962.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000448962.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000448962.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000460761.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000460761.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000460761.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000460761.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000460761.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000484121.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000484121.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000484121.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000484121.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000484121.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000484121.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000484121.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000484121.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000484121.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000495002.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000495002.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000495002.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000495002.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000495002.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000495002.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000495002.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000495002.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000495002.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000495002.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000626547.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000626547.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000626547.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000626547.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008840.hg.1 ENST00000626547.2 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000302907.8 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000302907.8 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000302907.8 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000302907.8 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000302907.8 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000302907.8 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000302907.8 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000302907.8 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000302907.8 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000302907.8 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000391751.7 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000391751.7 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000391751.7 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000391751.7 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000391751.7 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000391751.7 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000391751.7 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000391752.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000391752.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000391752.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000391752.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000391752.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000391752.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000391752.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000391752.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000391752.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000391753.6 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000391753.6 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000391753.6 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000391753.6 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000391753.6 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000391753.6 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000391753.6 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000391753.6 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000402367.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000402367.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000402367.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000402367.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000402367.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000402367.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000402367.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000402367.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000402367.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000402367.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000402367.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000402367.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000436445.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000436445.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000436445.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000436445.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000436445.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000436445.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000436445.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000441429.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000441429.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000441429.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000441429.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000441429.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000441429.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000441429.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000441429.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000441429.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000441429.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000441429.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000441429.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000445961.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000445961.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000445961.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000445961.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000445961.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000445961.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000445961.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000445961.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000445961.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000445961.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000448962.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000448962.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000448962.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000448962.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000448962.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000448962.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000460761.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000460761.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000460761.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000460761.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000460761.1 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000484121.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000484121.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000484121.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000484121.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000484121.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000484121.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000484121.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000484121.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000484121.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000495002.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000495002.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000495002.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000495002.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000495002.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000495002.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000495002.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000495002.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000495002.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000495002.5 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000626547.2 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000626547.2 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000626547.2 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000626547.2 Ensembl
Affymetrix Human Clariom D Assay TC1900008840.hg.1 ENST00000626547.2 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline