This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01591082_m1
See other TIAL1 GE Assays ›
Gene Symbol:
TIAL1
Gene Name:
TIA1 cytotoxic granule-associated RNA binding protein-like 1
Gene Aliases:
TCBP, TIAR
Chromosome Location:
Chr.10: 119573465 - 119597247 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01591082_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TIAL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001323968.1 NP_001310897.1 12 - 13 1668 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Apoptosis 3' Most Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
apoptotic process
defense response
germ cell development
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
stem cell division
nucleotide binding
RNA binding
AU-rich element binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0