This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01591985_g1
See other EEF1A1 GE Assays ›
Gene Symbol:
EEF1A1
Gene Name:
eukaryotic translation elongation factor 1 alpha 1
Gene Aliases:
CCS-3, CCS3, EE1A1, EEF-1, EEF1A, EF-Tu, EF1A, GRAF-1EF, HNGC:16303, LENG7, PTI1, eEF1A-1
Chromosome Location:
Chr.6: 73515750 - 73521032 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01591985_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EEF1A1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001402.5 NP_001393.1 4 - 5 685 144
XM_011535514.2 XP_011533816.1 4 - 5 1320 144
GenBank mRNA AF328730.1 - 1 393 144
AF397403.1 - 2 - 3 430 144
AJ420488.1 - 3 - 4 644 144
AK024888.1 - 1 683 144
AK026650.1 - 1 685 144
AK055058.1 - 4 - 5 685 144
AK058051.1 - 4 - 5 685 144
AK058163.1 - 3 - 4 503 144
AK098510.1 - 4 - 5 685 144
AK098518.1 - 4 - 5 545 144
AK098587.1 - 1 - 2 113 144
AK098623.1 - 4 - 5 685 144
AK127384.1 - 4 - 5 685 144
AK222515.1 - 1 685 144
AK222519.1 - 1 334 144
AK222523.1 - 1 685 144
AK222551.1 - 1 685 144
AK222982.1 - 1 685 144
AK223030.1 - 1 682 144
AK223042.1 - 1 540 144
AK223046.1 - 1 688 144
AK289374.1 - 4 - 5 685 144
AK289442.1 - 4 - 5 623 144
AK290691.1 - 4 - 5 685 144
AK292639.1 - 4 - 5 1001 144
AK300922.1 - 4 - 5 546 144
AK315827.1 - 4 - 5 676 144
AY043301.1 - 3 - 4 1032 144
BC008587.1 - 1 671 3447654 144
BC009733.1 - 1 635 3877516 144
BC009875.2 - 1 675 3948601 144
BC010735.1 - 1 664 3898364 144
BC012891.1 - 1 663 3858483 144
BC014224.2 - 1 674 4335756 144
BC014377.1 - 1 97 144
BC018150.1 - 1 671 3851939 144
BC018641.2 - 1 671 3660140 144
BC018850.2 - 1 674 144
BC019050.2 - 1 637 144
BC020477.2 - 1 334 144
BC021686.1 - 1 690 3930863 144
BC028674.1 - 1 689 4107346 144
BC029337.1 - 1 659 144
BC029343.1 - 1 657 144
BC029997.1 - 1 672 144
BC035877.1 - 1 686 144
BC038339.1 - 1 671 3461588 144
BC057391.1 - 1 640 4339606 144
BC063511.1 - 1 108 144
BC066893.1 - 1 685 4818863 144
BC071619.1 - 2 601 4448926 144
BC071727.1 - 1 640 4514479 144
BC071741.1 - 1 665 3885539 144
BC071841.1 - 1 660 4587981 144
BC072385.1 - 1 672 5585092 144
BC082268.1 - 1 664 6729595 144
BC094687.1 - 2 577 30366186 144
BC111051.1 - 1 675 5585079 144
EF219140.1 - 4 - 5 845 144
EF362804.1 - 4 - 5 915 144
M29548.1 - 1 - 2 217 144
X03558.1 - 4 - 5 675 144
X16869.1 - 3 - 4 637 144

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600012296.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06001875.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EEF1A1

Panther Classification:

Molecular Function -

translation factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
translational elongation
cellular response to epidermal growth factor stimulus
regulation of chaperone-mediated autophagy
tRNA binding
translation elongation factor activity
GTPase activity
protein binding
GTP binding
protein kinase binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0