This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01592085_m1
See other TMPRSS11F GE Assays ›
Gene Symbol:
TMPRSS11F
Gene Name:
transmembrane protease, serine 11F
Gene Aliases:
-
Chromosome Location:
Chr.4: 68053198 - 68129869 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01592085_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMPRSS11F
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_207407.2 NP_997290.2 7 - 8 803 90
GenBank mRNA AK122625.1 - 7 - 8 803 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400010898.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04001255.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Protease Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TMPRSS11F

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
serine-type endopeptidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0400010898.hg.1 ENST00000356291.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010898.hg.1 ENST00000356291.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010898.hg.1 ENST00000356291.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010898.hg.1 ENST00000356291.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010898.hg.1 ENST00000356291.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010898.hg.1 ENST00000356291.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010898.hg.1 ENST00000356291.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010898.hg.1 ENST00000356291.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010898.hg.1 ENST00000356291.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010898.hg.1 ENST00000356291.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010898.hg.1 ENST00000356291.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0400010898.hg.1 NM_207407.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010898.hg.1 NM_207407.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010898.hg.1 NM_207407.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010898.hg.1 NM_207407.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010898.hg.1 NM_207407.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010898.hg.1 NM_207407.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010898.hg.1 NM_207407.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010898.hg.1 NM_207407.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010898.hg.1 NM_207407.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010898.hg.1 NM_207407.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010898.hg.1 NM_207407.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04017078.hg.1 TC04001255.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04017078.hg.1 ENST00000356291.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04017078.hg.1 NM_207407.2 Refseq
Affymetrix Human Clariom D Assay TC0400010898.hg.1 ENST00000356291.2 Ensembl
Affymetrix Human Clariom D Assay TC0400010898.hg.1 ENST00000356291.2 Ensembl
Affymetrix Human Clariom D Assay TC0400010898.hg.1 ENST00000356291.2 Ensembl
Affymetrix Human Clariom D Assay TC0400010898.hg.1 ENST00000356291.2 Ensembl
Affymetrix Human Clariom D Assay TC0400010898.hg.1 ENST00000356291.2 Ensembl
Affymetrix Human Clariom D Assay TC0400010898.hg.1 ENST00000356291.2 Ensembl
Affymetrix Human Clariom D Assay TC0400010898.hg.1 ENST00000356291.2 Ensembl
Affymetrix Human Clariom D Assay TC0400010898.hg.1 ENST00000356291.2 Ensembl
Affymetrix Human Clariom D Assay TC0400010898.hg.1 ENST00000356291.2 Ensembl
Affymetrix Human Clariom D Assay TC0400010898.hg.1 ENST00000356291.2 Ensembl
Affymetrix Human Clariom D Assay TC0400010898.hg.1 ENST00000356291.2 Ensembl
Affymetrix Human Clariom D Assay TC0400010898.hg.1 NM_207407.2 Refseq
Affymetrix Human Clariom D Assay TC0400010898.hg.1 NM_207407.2 Refseq
Affymetrix Human Clariom D Assay TC0400010898.hg.1 NM_207407.2 Refseq
Affymetrix Human Clariom D Assay TC0400010898.hg.1 NM_207407.2 Refseq
Affymetrix Human Clariom D Assay TC0400010898.hg.1 NM_207407.2 Refseq
Affymetrix Human Clariom D Assay TC0400010898.hg.1 NM_207407.2 Refseq
Affymetrix Human Clariom D Assay TC0400010898.hg.1 NM_207407.2 Refseq
Affymetrix Human Clariom D Assay TC0400010898.hg.1 NM_207407.2 Refseq
Affymetrix Human Clariom D Assay TC0400010898.hg.1 NM_207407.2 Refseq
Affymetrix Human Clariom D Assay TC0400010898.hg.1 NM_207407.2 Refseq
Affymetrix Human Clariom D Assay TC0400010898.hg.1 NM_207407.2 Refseq
Affymetrix Human Clariom D Assay PSR0400174100.hg.1 ENST00000356291.2 Ensembl
Affymetrix Human Clariom D Assay PSR0400174100.hg.1 NM_207407.2 Refseq
Affymetrix Human Clariom D Assay PSR0400174100.hg.1 ENST00000356291.2 Ensembl
Affymetrix Human Clariom D Assay PSR0400174100.hg.1 NM_207407.2 Refseq
Affymetrix Human Clariom D Assay PSR0400174100.hg.1 TC0400010898.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0