This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01592442_m1
See other NAT6 GE Assays ›
Gene Symbol:
NAT6
Gene Name:
N-acetyltransferase 6
Gene Aliases:
FUS-2, FUS2
Chromosome Location:
Chr.3: 50296402 - 50299421 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01592442_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NAT6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001200016.1 NP_001186945.1 1 - 2 213 140
GenBank mRNA AK313803.1 - 1 - 2 213 140

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300013983.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03003412.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK313803.1,NM_001200016.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Process(es)

N-acetyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000354862.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000354862.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000354862.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000354862.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000354862.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000354862.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000354862.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000354862.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000354862.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000417393.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000417393.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000417393.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000417393.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000417393.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000417393.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000417393.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000417393.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000442620.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000442620.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000442620.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000442620.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000442620.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000442620.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000442620.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000442620.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000443094.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000443094.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000443094.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000443094.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000443094.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000443094.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000443094.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000443094.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000443094.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000443094.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000443842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000443842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000443842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000443842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000443842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000443842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000443842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000443842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000443842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000443842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000443842.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000450489.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000450489.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000450489.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000450489.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000450489.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000450489.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000450489.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000450489.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000452674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000452674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000452674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000452674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000452674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 ENST00000452674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 NM_001200016.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 NM_001200016.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 NM_001200016.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 NM_001200016.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 NM_001200016.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 NM_001200016.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 NM_001200016.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 NM_001200016.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 NM_001200016.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 NM_001200016.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 NM_001200018.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 NM_001200018.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 NM_001200018.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 NM_001200018.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 NM_001200018.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 NM_001200018.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 NM_001200018.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 NM_001200018.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 NM_001200018.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 NM_012191.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 NM_012191.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 NM_012191.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 NM_012191.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 NM_012191.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 NM_012191.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 NM_012191.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 NM_012191.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300013983.hg.1 NM_012191.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0