This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01594825_m1
See other LOC400794 GE Assays ›
Gene Symbol:
LOC400794
Gene Name:
uncharacterized LOC400794
Gene Aliases:
-
Chromosome Location:
Chr.1: 165476842 - 165582155 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01594825_m1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 360.22
360.22
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC400794
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_026744.2 2 - 3 886 82
GenBank mRNA AK093132.1 - 2 - 3 835 82
BC030596.2 - 2 - 3 859 82

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream leucine rich repeat containing 52 LRRC52 -10.1kb Anti-sense Chr. 1 - 165544241 - 165563948 on NCBI Build GRCh38
Downstream microsomal glutathione S-transferase 3 MGST3 GST-III 55.5kb Anti-sense Chr. 1 - 165630873 - 165656136 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100016256.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01003465.hg.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000415000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000415000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000415000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000415000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000418925.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000418925.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000418925.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000454251.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000454251.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 ENST00000454251.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01053942.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01053942.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01053942.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01053942.hg.1 NR_026744.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01053942.hg.1 TC01003465.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000415000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000415000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000415000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000415000.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000418925.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000418925.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000418925.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000452283.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000454251.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000454251.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 ENST00000454251.1 Ensembl
Affymetrix Human Clariom D Assay TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Clariom D Assay TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Clariom D Assay TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Clariom D Assay TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Clariom D Assay TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Clariom D Assay TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Clariom D Assay TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Clariom D Assay TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Clariom D Assay TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Clariom D Assay TC0100016256.hg.1 NR_026744.2 Refseq
Affymetrix Human Clariom D Assay PSR0100223743.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100223743.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100223743.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100223743.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100223743.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100223743.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100223743.hg.1 NR_026744.2 Refseq
Affymetrix Human Clariom D Assay PSR0100223743.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100223743.hg.1 ENST00000416424.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100223743.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100223743.hg.1 ENST00000421273.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100223743.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100223743.hg.1 ENST00000438275.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100223743.hg.1 NR_026744.2 Refseq
Affymetrix Human Clariom D Assay PSR0100223743.hg.1 TC0100016256.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-p58g2:80/100.66.72.198:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0