Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000300128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000300128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000300128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000300128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000300128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000300128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000300128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000300128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000300128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000300128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000300128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000300128.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000379391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000379391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000379391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000379391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000379391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000379391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000379391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000379391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000379391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000379391.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000553654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000553654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000553654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000553654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000554340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000554340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000554340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000554340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000554340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000554340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000554340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000554340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
ENST00000554340.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
NM_001130963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
NM_001130963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
NM_001130963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
NM_001130963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
NM_001130963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
NM_001130963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
NM_001130963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
NM_001130963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
NM_001130963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
NM_001130963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
NM_001130963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
NM_001130963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
NM_015257.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
NM_015257.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
NM_015257.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
NM_015257.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
NM_015257.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
NM_015257.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
NM_015257.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
NM_015257.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
NM_015257.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200010946.hg.1 |
NM_015257.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021761.hg.1 |
ENST00000300128.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021761.hg.1 |
ENST00000379391.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021761.hg.1 |
NM_001130963.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021761.hg.1 |
NM_015257.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12021761.hg.1 |
TC12001619.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200184848.hg.1 |
ENST00000300128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200184848.hg.1 |
ENST00000379391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200184848.hg.1 |
NM_001130963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200184848.hg.1 |
NM_015257.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200184848.hg.1 |
ENST00000300128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200184848.hg.1 |
ENST00000379391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200184848.hg.1 |
NM_001130963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200184848.hg.1 |
NM_015257.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200184848.hg.1 |
TC1200010946.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000300128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000300128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000300128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000300128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000300128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000300128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000300128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000300128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000300128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000300128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000300128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000300128.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000379391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000379391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000379391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000379391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000379391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000379391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000379391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000379391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000379391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000379391.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000553654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000553654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000553654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000553654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000554340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000554340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000554340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000554340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000554340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000554340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000554340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000554340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
ENST00000554340.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
NM_001130963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
NM_001130963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
NM_001130963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
NM_001130963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
NM_001130963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
NM_001130963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
NM_001130963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
NM_001130963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
NM_001130963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
NM_001130963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
NM_001130963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
NM_001130963.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
NM_015257.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
NM_015257.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
NM_015257.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
NM_015257.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
NM_015257.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
NM_015257.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
NM_015257.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
NM_015257.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
NM_015257.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200010946.hg.1 |
NM_015257.2 |
Refseq |