This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01594999_m1
See other RBM41 GE Assays ›
Gene Symbol:
RBM41
Gene Name:
RNA binding motif protein 41
Gene Aliases:
-
Chromosome Location:
Chr.X: 107061885 - 107118827 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01594999_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBM41
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001324242.1 NP_001311171.1 5 - 6 646 130
NM_001324243.1 NP_001311172.1 5 - 6 739 130
NM_001324244.1 NP_001311173.1 5 - 6 646 130
NR_136730.1 5 - 6 646 130
NR_136731.1 5 - 6 739 130
XM_011530980.2 XP_011529282.1 5 - 6 738 130
GenBank mRNA AK023732.1 - 5 - 6 642 130

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
developmental process
nucleotide binding
protein binding
U12 snRNA binding
pre-mRNA intronic binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0