This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01595000_m1
See other RBM41 GE Assays ›
Gene Symbol:
RBM41
Gene Name:
RNA binding motif protein 41
Gene Aliases:
-
Chromosome Location:
Chr.X: 107061885 - 107118827 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01595000_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBM41
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_136730.1 6 - 7 753 145
GenBank mRNA AK023732.1 - 6 - 7 749 145

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
developmental process
nucleotide binding
protein binding
U12 snRNA binding
pre-mRNA intronic binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000372479.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000372479.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000372479.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000372479.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000372479.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000372479.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000372479.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000372479.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000372479.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000372479.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000372479.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000372479.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000372479.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000372479.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000372487.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000372487.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000372487.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000372487.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000372487.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000372487.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000372487.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000372487.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000372487.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000372487.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000372487.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000372487.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000372487.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000434854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000434854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000434854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000434854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000434854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000434854.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000471079.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000471079.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000471079.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000471079.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000471079.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000474499.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000474499.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000475556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000475556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000475556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000475556.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000485676.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000485676.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000485676.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000485676.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000485676.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000485676.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000485676.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000485676.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_001171080.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_001171080.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_001171080.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_001171080.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_001171080.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_001171080.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_001171080.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_001171080.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_001171080.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_001171080.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_001171080.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_001171080.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_001171080.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_001171080.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000372479.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000372479.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000372479.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000372479.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000372479.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000372479.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000372479.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000372479.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000372479.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000372479.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000372479.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000372479.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000372479.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000372479.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000372487.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000372487.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000372487.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000372487.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000372487.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000372487.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000372487.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000372487.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000372487.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000372487.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000372487.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000372487.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000372487.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000434854.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000434854.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000434854.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000434854.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000434854.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000434854.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000471079.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000471079.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000471079.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000471079.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000471079.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000474499.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000474499.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000475556.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000475556.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000475556.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000475556.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000485676.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000485676.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000485676.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000485676.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000485676.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000485676.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000485676.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000485676.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 ENST00000495517.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_001171080.1 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_001171080.1 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_001171080.1 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_001171080.1 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_001171080.1 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_001171080.1 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_001171080.1 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_001171080.1 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_001171080.1 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_001171080.1 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_001171080.1 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_001171080.1 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_001171080.1 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_001171080.1 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_018301.4 Refseq
Affymetrix Human Clariom D Assay TC0X00010462.hg.1 NM_018301.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0