This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01596840_g1
See other RBPMS2 GE Assays ›
Gene Symbol:
RBPMS2
Gene Name:
RNA binding protein with multiple splicing 2
Gene Aliases:
-
Chromosome Location:
Chr.15: 64739894 - 64775571 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01596840_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBPMS2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_194272.1 NP_919248.1 6 - 7 822 156
GenBank mRNA AK124123.1 - 6 - 7 570 156
AK127873.1 - 6 - 7 838 156
AY369207.1 - 6 - 7 822 156
BC131612.1 - 5 - 6 474 156
BC148208.1 - 5 - 6 474 156

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1500009756.hg.1
This assay targets the same location as HTA transcript cluster(s) TC15001553.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RBPMS2

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of BMP signaling pathway
embryonic digestive tract morphogenesis
positive regulation of smooth muscle cell proliferation
negative regulation of smooth muscle cell differentiation
nucleotide binding
mRNA binding
protein binding
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR15014762.hg.1 ENST00000300069.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15014762.hg.1 ENST00000560606.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR15014762.hg.1 NM_194272.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR15014762.hg.1 TC15001553.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000300069.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000300069.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000300069.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000300069.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000300069.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000300069.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000300069.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000300069.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000300069.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000560606.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000560606.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000560606.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000560606.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000560606.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000560606.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000560606.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 ENST00000560606.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 NM_194272.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 NM_194272.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 NM_194272.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 NM_194272.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 NM_194272.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 NM_194272.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 NM_194272.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 NM_194272.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500009756.hg.1 NM_194272.2 Refseq
Affymetrix Human Clariom D Assay PSR1500172069.hg.1 ENST00000300069.4 Ensembl
Affymetrix Human Clariom D Assay PSR1500172069.hg.1 ENST00000560606.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500172069.hg.1 NM_194272.2 Refseq
Affymetrix Human Clariom D Assay PSR1500172069.hg.1 ENST00000300069.4 Ensembl
Affymetrix Human Clariom D Assay PSR1500172069.hg.1 ENST00000560606.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500172069.hg.1 NM_194272.2 Refseq
Affymetrix Human Clariom D Assay PSR1500172069.hg.1 TC1500009756.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000300069.4 Ensembl
Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000300069.4 Ensembl
Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000300069.4 Ensembl
Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000300069.4 Ensembl
Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000300069.4 Ensembl
Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000300069.4 Ensembl
Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000300069.4 Ensembl
Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000300069.4 Ensembl
Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000300069.4 Ensembl
Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000560606.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000560606.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000560606.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000560606.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000560606.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000560606.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000560606.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009756.hg.1 ENST00000560606.5 Ensembl
Affymetrix Human Clariom D Assay TC1500009756.hg.1 NM_194272.2 Refseq
Affymetrix Human Clariom D Assay TC1500009756.hg.1 NM_194272.2 Refseq
Affymetrix Human Clariom D Assay TC1500009756.hg.1 NM_194272.2 Refseq
Affymetrix Human Clariom D Assay TC1500009756.hg.1 NM_194272.2 Refseq
Affymetrix Human Clariom D Assay TC1500009756.hg.1 NM_194272.2 Refseq
Affymetrix Human Clariom D Assay TC1500009756.hg.1 NM_194272.2 Refseq
Affymetrix Human Clariom D Assay TC1500009756.hg.1 NM_194272.2 Refseq
Affymetrix Human Clariom D Assay TC1500009756.hg.1 NM_194272.2 Refseq
Affymetrix Human Clariom D Assay TC1500009756.hg.1 NM_194272.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline